Skip to Content
Merck
All Photos(3)

Key Documents

733504

Sigma-Aldrich

Bismuth selenide

greener alternative

granular (melted), ≥99.995% trace metals basis

Synonym(s):

Dibismuth triselenide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi2Se3
CAS Number:
Molecular Weight:
654.84
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.995% trace metals basis

form

granular (melted)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

710 °C (lit.)

greener alternative category

SMILES string

[Se]=[Bi][Se][Bi]=[Se]

InChI

1S/2Bi.3Se

InChI key

OMEPJWROJCQMMU-UHFFFAOYSA-N

General description

Bismuth selenide (Bi2Se3), a thermoelectric material that acts as a topological insulator, exhibits helical spin polarization and strong spin-orbit coupling.
Bismuth selenide is a semiconductor thermoelectric material.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Bi2Se3 can be used in the formation of nanocomposites for antitumor thermo-chemotherapy and imaging. It can also be used in the fabrication of electronic devices such as field effect transistors (FETs) and photodetectors.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topological insulators: Oscillations in the ribbons.
Thomas Ihn
Nature materials, 9(3), 187-188 (2010-02-20)
Observation of inverse spin Hall effect in bismuth selenide
Deorani P, et al.
Physical Review. B, Condensed Matter and Materials Physics, 90(9), 094403-094403 (2014)
Insulating behavior in ultrathin bismuth selenide field effect transistors
Cho S, et al.
Nano Letters, 11(5), 1925-1927 (2011)
Multifunctional bismuth selenide nanocomposites for antitumor thermo-chemotherapy and imaging
Li Z, et al.
ACS Nano, 10(1), 984-997 (2016)
High performance broadband photodetector using fabricated nanowires of bismuth selenide
Sharma A, et al.
Scientific reports, 6(1), 19138-19138 (2016)

Articles

Thermoelectric materials comprise a wide range of solid compounds distinguished by their ability to convert thermal and electrical energy.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

Higher transition metal silicides are ideal for anisotropic thermoelectric conversion due to their Seebeck coefficient anisotropy and mechanical properties.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service