Skip to Content
Merck
All Photos(1)

Documents

727989

Sigma-Aldrich

Ethyldimethylpropylammonium bis(trifluoromethylsulfonyl)imide

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18F6N2O4S2
CAS Number:
Molecular Weight:
396.37
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

impurities

≤1.0% water

SMILES string

CCC[N+](C)(C)CC.FC(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

InChI

1S/C7H18N.C2F6NO4S2/c1-5-7-8(3,4)6-2;3-1(4,5)14(10,11)9-15(12,13)2(6,7)8/h5-7H2,1-4H3;/q+1;-1

InChI key

FKXJTTMLNYZAOH-UHFFFAOYSA-N

General description

Ethyldimethylpropylammonium bis(trifluoromethylsulfonyl)imide is an ionic liquid with high dielectrical permittivity, ionic conductivity and low viscosity.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Confinement of 1-butyl-3-methylimidazolium nitrate in metallic silver
Neouze, Marie-Alexandra, and Marco Litschauer
The Journal of Physical Chemistry, 112.51, 16721-16725 (2008)
Parametrization of 1-butyl-3-methylimidazolium hexafluorophosphate/nitrate ionic liquid for the GROMOS force field
Micaelo, Nuno M., Antonio M. Baptista, and Claudio M. Soares
Analytical Chemistry, 110.29, 14444-14451 (2006)
Electrical double layer capacitors with sucrose derived carbon electrodes in ionic liquid electrolytes.
Wei L and Yushin G.
Journal of Power Sources, 196(8), 4072-4079 (2011)
Thermochemical properties of 1-butyl-3-methylimidazolium nitrate
Strechan, A. A., et al.
Electrochimica Acta, 474.1, 25-31 (2008)
Density and viscosity of 1-butyl-3-methylimidazolium nitrate with ethanol, 1-propanol, or 1-butanol at several temperatures
Mokhtarani, Babak, et al.
Analytical Chemistry, 41.12, 1432-1438 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service