Skip to Content
Merck
All Photos(1)

Key Documents

701920

Sigma-Aldrich

2-((Di-tert-butylphosphinomethyl)-6-diethylaminomethyl)pyridine

Synonym(s):

PNN

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H35N2P
CAS Number:
Molecular Weight:
322.47
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand

refractive index

n20/D 1.519

density

0.936 g/mL at 25 °C

functional group

phosphine

SMILES string

CCN(CC)Cc1cccc(CP(C(C)(C)C)C(C)(C)C)n1

InChI

1S/C19H35N2P/c1-9-21(10-2)14-16-12-11-13-17(20-16)15-22(18(3,4)5)19(6,7)8/h11-13H,9-10,14-15H2,1-8H3

InChI key

MTBWGMDPQBSPGF-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3 - Water-react 3

Target Organs

Respiratory system

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

154.0 °F

Flash Point(C)

67.8 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsung-Hsun Hsieh et al.
Cerebral cortex (New York, N.Y. : 1991), 27(12), 5509-5524 (2016-12-03)
Many neuropsychiatric symptoms that follow traumatic brain injury (TBI), including mood disorders, sleep disturbance, chronic pain, and posttraumatic epilepsy (PTE) are attributable to compromised cortical inhibition. However, the temporal trajectory of cortical inhibition loss and its underlying mechanisms are not
Peyvand Ghaderyan et al.
International journal of psychophysiology : official journal of the International Organization of Psychophysiology, 110, 91-101 (2016-10-27)
Automatic workload estimation has received much attention because of its application in error prevention, diagnosis, and treatment of neural system impairment. The development of a simple but reliable method using minimum number of psychophysiological signals is a challenge in automatic
Weijian Cong et al.
Biomedical engineering online, 16(1), 124-124 (2017-11-01)
3D ultrasound volume reconstruction from B-model ultrasound slices can provide more clearly and intuitive structure of tissue and lesion for the clinician. This paper proposes a novel Global Path Matching method for the 3D reconstruction of freehand ultrasound images. The
Malathy Emperuman et al.
Sensors (Basel, Switzerland), 20(3) (2020-02-06)
Sensor devices in wireless sensor networks are vulnerable to faults during their operation in unmonitored and hazardous environments. Though various methods have been proposed by researchers to detect sensor faults, only very few research studies have reported on capturing the
Xiaoyun Xu et al.
Experimental neurology, 303, 59-71 (2018-02-10)
The partial recovery that can occur after a stroke has been attributed to structural and functional plasticity that compensate for damage and lost functions. This plasticity is thought to be limited in part by the presence of growth inhibitors in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service