Skip to Content
Merck
All Photos(1)

Key Documents

701734

Sigma-Aldrich

Bromobenzene-d5

reagent grade, ≥99 atom % D, ≥99% (CP)

Synonym(s):

Pentadeuterobromobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6D5Br
CAS Number:
Molecular Weight:
162.04
Beilstein:
1866602
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

grade

reagent grade

isotopic purity

≥99 atom % D

Assay

≥99% (CP)

form

liquid

refractive index

n20/D 1.5585 (lit.)

bp

156 °C
53 °C/23 mmHg (lit.)

density

1.538 g/mL at 25 °C
1.539 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

mass shift

M+5

SMILES string

[2H]c1c([2H])c([2H])c(Br)c([2H])c1[2H]

InChI

1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D

InChI key

QARVLSVVCXYDNA-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Calixarenes. 5. Dynamic NMR characteristics of p-tert-butylcalix [4]-arene and p-tert-butylcalix [8] arene.
Gutsche CD and Bauer LJ.
Tetrahedron Letters, 22(48), 4763-4766 (1981)
Optical constants, and vibrational assignment of liquid bromobenzene-d5 between 4000 and 400cm-1 at 25? C.
Keefe CD and Donovan LA.
Journal of Molecular Structure, 597(1), 259-268 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service