Skip to Content
Merck
All Photos(5)

Key Documents

701602

Sigma-Aldrich

[1,1′-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II)

greener alternative

98%

Synonym(s):

PdCl2(dtbpf)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H44Cl2FeP2Pd
CAS Number:
Molecular Weight:
651.75
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

powder

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

203-208 °C

greener alternative category

storage temp.

−20°C

SMILES string

[Fe].Cl[Pd]Cl.CC(C)(C)P([C]1[CH][CH][CH][CH]1)C(C)(C)C.CC(C)(C)P([C]2[CH][CH][CH][CH]2)C(C)(C)C

InChI

1S/2C13H22P.2ClH.Fe.Pd/c2*1-12(2,3)14(13(4,5)6)11-9-7-8-10-11;;;;/h2*7-10H,1-6H3;2*1H;;/q;;;;;+2/p-2

InChI key

JQZFOBWXNREQLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Find details here.
[1,1′-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II) (PdCl2(dtbpf)) participates as catalyst in Suzuki and Heck reactions. It is a preformed catalyst synthesized by a group of researchers at Johnson Matthey′s Catalysis and Chiral Technologies. It is an active catalyst for various Pd catalyzed cross-coupling reactions.
[1,1′-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II) is an air-stable, highly active and versatile catalyst. It is an efficient catalyst for cross-coupling reactions. It participates as catalyst in various coupling reactions of the fine chemicals and pharmaceutical chemicals. It is commonly employed as homogeneous catalyst in the Suzuki reactions.

PdCl2(dtbpf) is an air-stable cross-coupling catalyst used in the Suzuki coupling of various aryl chlorides.

Application

[1,1′-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II) (PdCl2(dtbpf)) may be used as catalyst in the Suzuki cross-coupling reaction of various aromatic and heteroaromatic halides with methyliminodiacetic acid derivatives (MIDA boronates) at room temperature (in the absence of organic solvent). It may be employed as catalyst for the synthesis of chalcones, via cross-coupling reactions.
[1,1′-Bis(di-tert-butylphosphino)ferrocene]dichloropalladium(II) was used in the preparation of [PdCl2(bisphosphinometallocene)] complexes. These complexes are reported to be the catalyst precursors for the Buchwald-Hartwig reaction.
It is also employed as catalyst for greener Suzuki cross-coupling in TPGS-750-M.

On the Way Towards Greener Transition-Metal-Catalyzed Processes as Quantified by E Factors

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An Effective Activation of Palladium Phosphine Complexes in Aqueous Phase Reactions of Hetero-Aromatic Boronic Acids with Aryl Halides.
Bumagin NA, et al.
Chemistry of Heterocyclic Compounds, 50(1), 19-25 (2011)
Bruce H Lipshutz et al.
Organic letters, 10(7), 1329-1332 (2008-03-14)
A nonionic amphiphile such as Triton X-100 or the vitamin E-based PTS, both of which form nanomicelles in water, promotes Heck cross-couplings of non-water-soluble partners at ambient temperatures. These are the first examples of Heck reactions conducted in water (as
Gabriela A Grasa et al.
Organic letters, 9(26), 5489-5492 (2007-11-23)
alpha-Arylation of various ketones with aryl chlorides and bromides using the well-defined and air-stable (DtBPF)PdX2 (X = Cl, Br) catalysts gave 80-100% yield of the coupled products under relatively mild conditions at low catalyst loadings. The X-ray structure of (DtBPF)PdCl2
?-Arylation of ketones using highly active, air-stable (DtBPF) PdX2 (X= Cl, Br) catalysts
Grasa GA and Colacot TJ
Organic Letters, 9(26), 5489-5492 (2007)
CRC International Symposium: Cross Coupling and Organometallics.
Colacot T.
Platinum Metals Rev., 52(3), 172-172 (2008)

Articles

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

A variety of palladium-catalyzed cross-coupling reactions can be run under room temperature conditions in water with TPGS- 750-M, using a variety of commercially available palladium complexes and ligands.

Protocols

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

TPGS-750-M surfactant enables various reactions in water at room temperature, enhancing efficiency and versatility in synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service