Skip to Content
Merck
All Photos(1)

Documents

693049

Sigma-Aldrich

(R)-Tol-BINAP

Synonym(s):

(R)-(+)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H40P2
CAS Number:
Molecular Weight:
678.78
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

optical activity

[α]20/D +162°, c = 0.5 in benzene

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

mp

254-258 °C

functional group

phosphine

SMILES string

Cc1ccc(cc1)P(c2ccc(C)cc2)c3ccc4ccccc4c3-c5c(ccc6ccccc56)P(c7ccc(C)cc7)c8ccc(C)cc8

InChI

1S/C48H40P2/c1-33-13-23-39(24-14-33)49(40-25-15-34(2)16-26-40)45-31-21-37-9-5-7-11-43(37)47(45)48-44-12-8-6-10-38(44)22-32-46(48)50(41-27-17-35(3)18-28-41)42-29-19-36(4)20-30-42/h5-32H,1-4H3

InChI key

IOPQYDKQISFMJI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

BINAP is based on a bis naphthalene backbone with different phosphine derivatives. 2,2′-bis(di-p-tolylphosphino)-1,1′-binaphthyl (p-Tol-BINAP)·AgF complex catalyzed asymmetric Mukaiyama-type aldol reaction is reported.

Application

(R)-T-BINAP complexes derived from rhodium precursors are used for the asymmetric hydroformylation of vinyl acetate. It may be employed as chiral catalyst for allylation of N-tosyl α-imino esters.
Catalyst involved in studies of the role of excess phosphine ligand in enantioselective conjugate addition of ethylmagnesium bromide and α,β-unsaturated ester

Reactant serving as a precursor for:
  • Catalysts used for reductive amination of ketones
  • Rh(I)-catalyst for hydrogenation of acetamidoacrylic acid derivatives
  • Chiral platinum catalysts for asymmetric Baeyer-Villiger oxidation of cyclic ketones
  • CuI-Tol-BINAP catalysts for enantioselective Michael reactions of Grignard reagents to unsaturated esters
  • BINAP Pt Dications for cation trapping

Legal Information

Sold in collaboration with Takasago for research purposes only.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catalytic Asymmetric Aldol Reaction of Trimethoxysilyl Enol Ethers Using 2, 2'-Bis (di-p-tolylphosphino)-1, 1'-binaphthyl AgF Complex.
Yanagisawa A, et al.
Bulletin of the Chemical Society of Japan, 74(8), 1477-1484 (2001)
Asymmetric hydroformylation of vinyl acetate with BINAP-rhodium (I) complexes.
Hoegaerts D and Jacobs PA.
Tetrahedron Asymmetry, 10(15), 3039-3043 (1999)
Xiangming Fang et al.
The Journal of organic chemistry, 64(13), 4844-4849 (2001-10-25)
A new catalytic enantioselective approach for the formation of allyl alpha-amino acid derivatives by reaction of N-tosyl alpha-imino esters with allyl stannanes and silanes catalyzed by chiral copper(I) complexes has been developed. A series of different BINAP and phosphine-oxazoline (P,N)

Articles

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

The Baeyer-Villiger oxidation is the oxidative cleavage of a carbon-carbon bond adjacent to a carbonyl, which converts the ketones to esters and the cyclic ketones to lactones.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service