
(R)-Tol-BINAP

Synonym(s):
(R)-(+)-2,2′-Bis(di-p-tolylphosphino)-1,1′-binaphthyl
Empirical Formula (Hill Notation):
C48H40P2
CAS Number:
Molecular Weight:
678.78
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing