Skip to Content
Merck
All Photos(2)

Key Documents

677450

Sigma-Aldrich

Zinc oxide

nanopowder, <50 nm particle size (BET), >97%

Synonym(s):

Zinc monoxide, Zinc white

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnO
CAS Number:
Molecular Weight:
81.39
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

>97%

form

nanopowder

contains

6% Al as dopant

reaction suitability

reagent type: catalyst
core: zinc

surface area

>10.8 m2/g

particle size

<50 nm (BET)

SMILES string

O=[Zn]

InChI

1S/O.Zn

InChI key

XLOMVQKBTHCTTD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Employed in the preparation of NaZnSiO3OH, a novel chiral framework material which has potential application in ion exchange, adsorption or catalysis.

Legal Information

Product of Engi-Mat Co.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transparent Semiconducting ZnO:Al Thin Films Prepared by Spray Pyrolysis
W. T. Seeber, et al
Materials Science in Semiconductor Processing, 2, 45-55 (1999)
"Nanopowder synthesis of zinc oxide via solochemical processing.
Vaezi MR and Sadrnezhaad SK
Materials & Design, 28(2), 515-519 (2007)
Effect of Microstructural Control on Thermoelectric Properties of Hot-Pressed Aluminum-Doped Zinc Oxide
Y. Fujishiro, etal.
Journal of the American Ceramic Society. American Ceramic Society, 86, 2063-2066 (2003)
Efficient hybrid solar cells from zinc oxide nanoparticles and a conjugated polymer.
Beek WJE, et al.
Advanced Materials, 16(12), 1009-1013 (2004)
Air-stable inverted flexible polymer solar cells using zinc oxide nanoparticles as an electron selective layer.
Hau S, et al.
Applied Physics Letters, 92(25), 253301-253301 (2008)

Articles

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

Operation principle and market dominance of single crystalline silicon solar cells.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service