Skip to Content
AF
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Zinc oxide
Synonym(s):
Zinc oxide
Linear Formula:
ZnO
CAS Number:
1314-13-2
Molecular Weight:
81.39
EC Number:
215-222-5
Products
Product Category
salts (4)
Brand
SAFC (1)
Sigma-Aldrich (7)
Supelco (3)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Puriss (2)
ACS reagent (1)
Puriss P.A. (1)
analytical standard (1)
certified reference material (1)
pharmaceutical primary standard (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-13 of 13
Compare
Product Number
Description
Pricing
205532
Zinc oxide,
ReagentPlus
®
, powder, <5 μm particle size, 99.9%
Expand
View Pricing
544906
Zinc oxide,
nanopowder, <100 nm particle size
Expand
View Pricing
96479
Zinc oxide,
puriss. p.a., ACS reagent, ≥99.0% (KT)
Expand
View Pricing
677450
Zinc oxide,
nanopowder, <50 nm particle size (BET), >97%
Expand
View Pricing
255750
Zinc oxide,
99.99% trace metals basis
Expand
View Pricing
14439
Zinc oxide,
puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Expand
View Pricing
1.08849
Zinc oxide,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
1724747
Zinc oxide,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
204951
Zinc oxide,
99.999% trace metals basis
Expand
View Pricing
773999
Zinc oxide,
nanowires, Length 1 um
Expand
View Pricing
1.08846
Zinc oxide,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, JP, USP
Expand
View Pricing
PHR2724
Zinc oxide,
Pharmaceutical Secondary Standard; Certified Reference Material, certified reference material, pharmaceutical secondary standard, pkg of 500 mg
Expand
View Pricing
93632
Zinc oxide,
analytical standard
Expand
View Pricing