Skip to Content
Merck
All Photos(1)

Key Documents

611360

Supelco

Urea-13C,15N2 solution

NMR reference standard, 0.1 M in DMSO-d6 (99.9 atom % D), NMR tube size 8 mm × 7 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH415N2O
Molecular Weight:
63.03
UNSPSC Code:
12142201
NACRES:
NA.24

grade

NMR reference standard
analytical standard

concentration

0.1 M in DMSO-d6 (99.9 atom % D)

technique(s)

NMR: suitable

NMR tube size

8 mm × 7 in.

suitability

suitable for NMR (reference standard)

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection

Quantity

8 mm O.D. tube contains 1.9 mL
Each pack contains 10 tubes..

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service