Skip to Content
Merck
All Photos(3)

Key Documents

410896

Sigma-Aldrich

2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone

98%

Synonym(s):

1-[4-(2-Hydroxyethoxy)phenyl]-2-hydroxy-2-methyl-1-propan-1-one, 2-Hydroxy-1-[4-(2-hydroxyethoxy)phenyl]-2-methyl-1-propanone, 2-Hydroxy-2-methyl-1-[4-(2-hydroxyethoxy)phenyl]propan-1-one, 4-(2-Hydroxyethoxy)phenyl 2-hydroxy-2-propyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH2OC6H4COC(CH3)2OH
CAS Number:
Molecular Weight:
224.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

mp

88-90 °C (lit.)

SMILES string

CC(C)(O)C(=O)c1ccc(OCCO)cc1

InChI

1S/C12H16O4/c1-12(2,15)11(14)9-3-5-10(6-4-9)16-8-7-13/h3-6,13,15H,7-8H2,1-2H3

InChI key

GJKGAPPUXSSCFI-UHFFFAOYSA-N

Application

2-Hydroxy-4′-(2-hydroxyethoxy)-2-methylpropiophenone can be used as a photo-initiator to synthesize:
  • Polyacrylamide-grafted chitosan nanoparticles by copolymerization of acrylamide and chitosan nanoparticles.
  • Hydrophobic polyurethane sponge through thiol–ene Click reaction.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

not determined

Flash Point(C)

not determined

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Zhao et al.
Acta biomaterialia, 49, 66-77 (2016-11-09)
Development of natural protein-based fibrous scaffolds with tunable physical properties and biocompatibility is highly desirable to construct three-dimensional (3D), fully cellularized scaffolds for wound healing. Herein, we demonstrated a simple and effective technique to construct electrospun 3D fibrous scaffolds for
Jiangya Ma et al.
Carbohydrate polymers, 151, 565-575 (2016-07-31)
In the present work, a new flocculant, polyacrylamide-grafted chitosan nanoparticles (NCS-g-PAM), was synthesized by the copolymerization of acrylamide (AM) and chitosan nanoparticle (NCS) under ultraviolet irradiation using 2-hydroxy-4'-(2-hydroxyethoxy)-2-methylpropiophenone as photo-initiator. The NCS was prepared by the ionic gelation between chitosan
Deepak Kumar et al.
Cell transplantation, 25(12), 2213-2220 (2016-07-28)
Nucleus pulposus (NP) tissue damage can induce detrimental mechanical strain on the biomechanical performance of intervertebral discs (IVDs), causing subsequent disc degeneration. A novel, photocurable, injectable, synthetic polymer hydrogel (pHEMA-co-APMA grafted with PAA) has already demonstrated success in encapsulating and
Rami El Assal et al.
Scientific reports, 6, 39144-39144 (2016-12-23)
Cancer cells have been increasingly grown in pharmaceutical research to understand tumorigenesis and develop new therapeutic drugs. Currently, cells are typically grown using two-dimensional (2-D) cell culture approaches, where the native tumor microenvironment is difficult to recapitulate. Thus, one of
Che-Ying Kuo et al.
Tissue engineering. Part A, 24(5-6), 527-535 (2017-07-21)
The goal of this work is to develop an innovative method that combines bioprinting and endoscopic imaging to repair tympanic membrane perforations (TMPs). TMPs are a serious health issue because they can lead to both conductive hearing loss and repeated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service