Skip to Content
Merck
All Photos(2)

Key Documents

570303

Sigma-Aldrich

Poly(ethylene glycol)-block-poly(ε−caprolactone) methyl ether

PEG average Mn ~5,000, PCL average Mn ~5,000

Synonym(s):

Poly(ethylene glycol) methyl ether-block-poly(ε−caprolactone)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

form

solid

mol wt

PCL average Mn ~5,000
PEG average Mn ~5,000
average Mn ~10,000 (total)

degradation timeframe

>12 months

mp

52.8-56.1 °C
54.2-55.9 °C (lit.)

Mw/Mn

<1.4

PDI

<1.4

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Biocompatible diblock copolymer

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Madalina Icriverzi et al.
Materials (Basel, Switzerland), 12(20) (2019-10-23)
The potential of mesenchymal stem cells (MSCs) for implantology and cell-based therapy represents one of the major ongoing research subjects within the last decades. In bone regeneration applications, the various environmental factors including bioactive compounds such as growth factors, chemicals

Articles

The development of drugs that target specific locations within the human body remains one of the greatest challenges in biomedicine today.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

Professor Robert K. Prud’homme introduces flash nanoprecipitation (FNP) for nanoparticle fabrication, which is a scalable, rapid mixing process for nanoparticle formulations.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service