Skip to Content
Merck
All Photos(1)

Key Documents

569593

Sigma-Aldrich

4,4′-Dimethyl-2,2′-dipyridyl

99.5%, purified by sublimation

Synonym(s):

2,2′-Bi(γ-picoline), 4,4′-Dimethyl-2,2′-bipyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
128995
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.5%

form

solid

purified by

sublimation

mp

169-174 °C (lit.)

SMILES string

Cc1ccnc(c1)-c2cc(C)ccn2

InChI

1S/C12H12N2/c1-9-3-5-13-11(7-9)12-8-10(2)4-6-14-12/h3-8H,1-2H3

InChI key

NBPGPQJFYXNFKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4,4′-Dimethyl-2,2′-dipyridyl (dmbipy) may be used as a ligand in the synthesis of :
  • carbonato-bridged triangular trinuclear compounds and tetranuclear hydroxo-bridged compounds
  • dmbipy-copper bridged complexes, which show variation in crystal geometries, thermal and magnetic properties

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

"Ferromagnetic trinuclear carbonato-bridged and tetranuclear hydroxo-bridged Cu (II) compounds with 4, 4'-dimethyl-2, 2'-bipyridine as ligand. X-ray structure, spectroscopy and magnetism"
Albada VAN, et al.
Inorgorganica Chimica Acta, 331(01), 208- 215 (2002)
"Crystal Chemistry of the 4,4`-Dimethyl-2,2`bipyridine/Copper Bromide System"
Willett.DR, et al
Inorganic Chemistry, 40(17), 4342-4352 (2001)
Maxime Mourer et al.
Bioorganic & medicinal chemistry, 20(6), 2035-2041 (2012-03-01)
Various polycharged calix[4]arenes were assayed as anti-mycobacterial agents against Mycobacterium tuberculosis, H(37)Rv strain. The sulfonate, carboxylate and phosphonate anionic species displayed no activity. Cationic derivatives integrating four aminoethyl groups at the upper rim and two 6,6'-dimethyl-2,2'-bipyridyl- or 4,4'-dimethyl-2,2'-bithiazolyl subunits at
Yang-Hui Luo et al.
Dalton transactions (Cambridge, England : 2003), 43(44), 16937-16942 (2014-10-11)
A new iron(II) complex based on the 4,4'-dimethyl-2,2'-bipyridine ligand [Fe(4,4'-dmbpy)3(ClO4)(SCN)·3H2O (1·3H2O)] has been prepared and characterized. Structural studies and Hirshfeld surface analysis for complex 1·3H2O at three different temperatures (300, 240 and 130 K) are described. The UV-vis absorption spectrum
Tommy Siu-Ming Tang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(30), 10729-10740 (2015-06-23)
The synthesis, characterization, photophysics, lipophilicity, and cellular properties of new phosphorescent ruthenium(II) polypyridine complexes functionalized with a dibenzocyclooctyne (DIBO) or amine moiety [Ru(N^N)2 (L)](PF6 )2 are reported (L=4-(13-N-(3,4:7,8-dibenzocyclooctyne-5-oxycarbonyl) amino-4,7,10-trioxa-tridecanyl-aminocarbonyl-oxy-methyl)-4'-methyl-2,2'-bipyridine bpy-DIBO, N^N=2,2'-bipyridine bpy (1 a), 1,10-phenanthroline phen (2 a); L=4-(13-amino-4,7,10-trioxa-tridecanylaminocarbonyl-oxy-methyl)-4'-methyl-2,2'-bipyridine bpy-NH2 , N^N=bpy

Articles

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

Arylboronic acids and esters, vital tools in chemical transformations, find extensive use, particularly in the Suzuki-Miyaura cross-coupling reaction.

See All

Protocols

We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.

We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service