Merck
  • Anti-mycobacterial activities of some cationic and anionic calix[4]arene derivatives.
Anti-mycobacterial activities of some cationic and anionic calix[4]arene derivatives.
Bioorganic & medicinal chemistry (2012-03-01)
Maxime Mourer, Hugues Massimba Dibama, Patricia Constant, Mamadou Daffé, Jean-Bernard Regnouf-de-Vains
ABSTRACT
Various polycharged calix[4]arenes were assayed as anti-mycobacterial agents against Mycobacterium tuberculosis, H(37)Rv strain. The sulfonate, carboxylate and phosphonate anionic species displayed no activity. Cationic derivatives integrating four aminoethyl groups at the upper rim and two 6,6'-dimethyl-2,2'-bipyridyl- or 4,4'-dimethyl-2,2'-bithiazolyl subunits at the lower rim were also found inactive against M. tuberculosis, while the unsubstituded and the 5,5'-dimethyl-2,2'-bipyridyl-analogues exhibited MIC values of 3.2 and 0.8μM respectively. Introduction of guanidinoethyl groups at the upper rim resulted, except for the 6,6'-dimethyl-2,2'-bipyridyl-derivative, in high anti-mycobacterial activities for the unsubstituted, the 5,5'-dimethyl-2,2'-bipyridyl- and the 4,4'-dimethyl-2,2'-bithiazolyl analogues, with MIC values of 0.8, 0.8 and 1.6μM, respectively, similar to those of current commercial anti-tuberculosis agents. The five more active substances were also evaluated against the isoniazid-resistant strain MYC5165, resulting in highly interesting micromolar or sub-micromolar MIC and IC(50), ca. 4-125 times more active than isoniazid. These preliminary results are attractive for the development of new anti-TB agents.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99.5%, purified by sublimation
Sigma-Aldrich
4,4′-Dimethyl-2,2′-dipyridyl, 99%
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
2,2′-Bipyridyl, ReagentPlus®, ≥99%
Supelco
Isoniazid, analytical standard, ≥99% (TLC)