Skip to Content
Merck
All Photos(4)

Key Documents

56485

Sigma-Aldrich

N-Hydroxysulfosuccinimide sodium salt

≥98% (HPLC), for peptide synthesis

Synonym(s):

Hydroxy-2,5-dioxopyrrolidine-3-sulfonicacid sodium salt, Sulfo-NHS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4NNaO6S
CAS Number:
Molecular Weight:
217.13
Beilstein:
6359129
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Product Name

N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)

Assay

≥98% (HPLC)

form

powder

composition

carbon, 21.57-22.68 (anhydrous)
nitrogen, 6.28-6.61 (anhydrous)

reaction suitability

reaction type: Addition Reactions

impurities

≤4% water

application(s)

peptide synthesis

functional group

imide
sulfonic acid

SMILES string

[Na+].ON1C(=O)CC(C1=O)S([O-])(=O)=O

InChI

1S/C4H5NO6S.Na/c6-3-1-2(12(9,10)11)4(7)5(3)8;/h2,8H,1H2,(H,9,10,11);/q;+1/p-1

InChI key

RPENMORRBUTCPR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

N-Hydroxysulfosuccinimide sodium salt is a water-soluble sulfonated analog of N-hydroxysuccinimide. It can undergo a coupling reaction with carboxylic acids in the presence of carbodiimides such as 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide(EDC) to form hydrophilic N-hydroxysulfosuccinimide active esters. These esters are useful in protein cross-linking experiments.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Sulfo-NHS is used for preparing hydrophilic active esters, e.g. for crosslinking agents

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-hydroxysulfosuccinimide active esters: bis (N-hydroxysulfosuccinimide) esters of two dicarboxylic acids are hydrophilic, membrane-impermeant, protein cross-linkers.
Staros JV
Biochemistry, 21(17), 3950-3955 (1982)
R Maydelid Trujillo-Nolasco et al.
Materials science & engineering. C, Materials for biological applications, 103, 109766-109766 (2019-07-28)
Radiosynovectomy is a technique used to decrease inflammation of the synovial tissue by intraarticular injection of a β-emitting radionuclide, such as 177Lu, which is suitable for radiotherapy due to its decay characteristics. Drug-encapsulating nanoparticles based on poly lactic‑co‑glycolic acid (PLGA)
Karen S Cheung Tung Shing et al.
Scientific reports, 8(1), 12457-12457 (2018-08-22)
A direct interaction between the erythropoietin (EPOR) and the beta-common (βc) receptors to form an Innate Repair Receptor (IRR) is controversial. On one hand, studies have shown a functional link between EPOR and βc receptor in tissue protection while others
P S Anjaneyulu et al.
International journal of peptide and protein research, 30(1), 117-124 (1987-07-01)
N-Hydroxysulfosuccinimide esters are reactive functional groups employed in a variety of protein modification reagents, especially cross-linking reagents. For these compounds, hydrolysis is the most important reaction competing for reaction of the esters with nucleophilic groups in proteins. We have employed
Eloy Povedano et al.
Scientific reports, 8(1), 6418-6418 (2018-04-25)
This paper describes two different electrochemical affinity biosensing approaches for the simple, fast and bisulfite and PCR-free quantification of 5-methylated cytosines (5-mC) in DNA using the anti-5-mC antibody as biorecognition element. One of the biosensing approaches used the anti-5-mC as

Articles

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Collagen molecules play a critical role in tissue architecture and strength, and in cell-matrix interactions as insoluble ligands to regulate the diverse phenotypic activities of cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service