Skip to Content
Merck
All Photos(1)

Key Documents

558958

Sigma-Aldrich

4-Chlorobenzyl bromide

97%

Synonym(s):

α-Bromo-p-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 1-Chloro-4-bromomethylbenzene, p-Chlorobenzyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4CH2Br
CAS Number:
Molecular Weight:
205.48
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

48-52 °C (lit.)

functional group

bromo
chloro

SMILES string

Clc1ccc(CBr)cc1

InChI

1S/C7H6BrCl/c8-5-6-1-3-7(9)4-2-6/h1-4H,5H2

InChI key

KQNBRMUBPRGXSL-UHFFFAOYSA-N

General description

4-Chlorobenzyl bromide undergoes carbonylation in the presence of dimer of chloro(1,5-cyclooctadiene)rhodium(I) to yield the corresponding phenylacetic acid. It can be synthesized by reacting 4-chlorobenzyl alcohol with bromodimethylsulfonium bromide (BDMS) It can also be synthesized by refluxing a mixture of 4-chlorobenzaldehyde, chlorotrimethylsilane, 1,1,3,3-tetramethyldisiloxane and lithium bromide.

Application

4-Chlorobenzyl bromide may be used to synthesize 1-(4-chlorobenzyl)-2-(pyrrolidin-1-yl-methyl)-1H-benzimidazole dihydrochloride

Packaging

4-Chlorobenzyl bromide may be used to synthesize:
  • 1-[1-[2-[(4-chlorobenzyl)thio]phenyl]vinyl]-1H-imidazole
  • 6-[4-(4-chlorobenzyl)piperazinyl]chromane
  • N,N′-bis(4-chlorobenzyl)piperazine
  • N-(4-chlorobenzyl)piperazine

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gazi S and Ananthakrishnan R
Royal Society of Chemistry Advances, 2(20), 7781- 7787 (2012)
"Reduction of carbonyl compounds promoted by silicon hydrides under the influence of trimethylsilyl-based reagent"
Aizpurua.MJ, et al.
Canadian Journal of Chemistry, 64(12), 2342-2347 (1986)
6-(4-Benzylpiperazin-1-yl) benzodioxanes as selective ligands at cloned primate dopamine D 4 receptors.
Hodgetts KJ, et al.
Bioorganic & Medicinal Chemistry, 9(12), 3207-3213 (2001)
Synthesis and antifungal activity of new 1-vinylimidazoles.
Ogata M, et al.
Journal of Medicinal Chemistry, 30(8), 1348-1354 (1987)
Synthesis of phenylacetic acids under rhodium-catalyzed carbonylation conditions.
Giroux A, et al.
Tetrahedron Letters, 41(40), 7601-7604 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service