Skip to Content
Merck
All Photos(2)

Key Documents

555959

Sigma-Aldrich

Dichloroborane dioxane complex solution

3 M in methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H8O2 · BHCl2
CAS Number:
Molecular Weight:
170.83
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reagent type: reductant

Quality Level

concentration

3 M in methylene chloride

density

1.321 g/mL at 25 °C

functional group

ether

storage temp.

2-8°C

SMILES string

Cl[BH-](Cl)[O+]1CCOCC1

InChI

1S/C4H9BCl2O2/c6-5(7)9-3-1-8-2-4-9/h5H,1-4H2

InChI key

RCKKTUSYECYDLY-UHFFFAOYSA-N

Application

Can readily substitute for common hydroborating reagents such as BH3 · THF and BMS.
Reactant for:
  • Hydroboration reactions
  • Preparation of alkenyl- and alkylboronic acids

Legal Information

U.S. Pat. No. 6,008,414

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josyula, K. V. B. et al
Tetrahedron Letters, 44, 7789-7789 (2003)
J V Kanth et al.
The Journal of organic chemistry, 66(16), 5359-5365 (2001-08-04)
Several less volatile oxygen-containing Lewis bases, such as tert-butyl methyl ether, dioxane, anisole, ethyl acetate, beta-chloroethyl ether, and monoglyme, were examined as prospective mono- and dichloroborane carriers. Dioxane, ethyl acetate, and beta-chloroethyl ether form relatively stable boron trichloride adducts, but
Kanth, J. V. B.; Brown, H. C.
Organic Letters, 1, 315-315 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service