Skip to Content
Merck
All Photos(1)

Key Documents

255106

Sigma-Aldrich

Dibromoborane dimethyl sulfide complex

Synonym(s):

Boron dibromide methyl sulfide complex, Dibromo(dimethyl sulfide)(hydro)boron, Dibromo(dimethyl sulfide)borane, Dibromoborane compound with dimethylsulfane, Dibromoborane dimethyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S · BHBr2
CAS Number:
Molecular Weight:
233.76
Beilstein:
4449985
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reagent type: reductant

mp

30-35 °C (lit.)

functional group

thioether

SMILES string

CSC.BrBBr

InChI

1S/C2H6S.BBr2H/c1-3-2;2-1-3/h1-2H3;1H

InChI key

PGSRDLGPKTVELT-UHFFFAOYSA-N

Application

Reactant for:
  • Reductive elimination C-C bond forming reactions
  • Lithiation-borylation reaction with Hoppe′s lithiated carbamates
  • Preparation of boron analogues of uracil
  • Boronation reactions for formation of cyclic boronium salts
  • Hydroboration reactions
  • Ring opening of terminal epoxides

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service