Skip to Content
Merck
All Photos(1)

Documents

490296

Sigma-Aldrich

Methanol-1-d

98 atom % D

Synonym(s):

Methan-d1-ol, Methanol-d1 (methyl-d1), Methyl-d1 Alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CDH2OH
CAS Number:
Molecular Weight:
33.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

refractive index

n20/D 1.326 (lit.)

bp

65.4 °C (lit.)

density

0.814 g/mL at 25 °C

mass shift

M+1

SMILES string

[2H]CO

InChI

1S/CH4O/c1-2/h2H,1H3/i1D

InChI key

OKKJLVBELUTLKV-MICDWDOJSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anita M Sutedja et al.
Molecules (Basel, Switzerland), 25(11) (2020-05-31)
Although the intake of jack bean (Canavalia ensiformis (L.) DC.), an underutilized tropical legume, can potentially decrease the risk of several chronic diseases, not much effort has been directed at profiling the polyphenolics contained therein. Hence, this work aimed to
Young Taek Han et al.
Chemical & pharmaceutical bulletin, 63(11), 907-912 (2015-08-21)
Concise and efficient synthesis of the proposed structure of damaurone D is accomplished in five steps without protection-deprotection operations. The key feature of our synthesis includes a versatile aldol reaction of the benzofuranone, provided by selective α-halogenation and intramolecular O-alkylation.
Jisu Yang et al.
Journal of Asian natural products research, 22(10), 966-975 (2020-08-22)
Herein, we report a concise and efficient method for the synthesis of p-hydroxyphenyl caffeate, a novel natural caffeic acid ester derivative isolated from Wedelia trilobata. The key feature of this synthesis is the Verley-Doebner modification of the Knoevenagel condensation of
Ga Eun Han et al.
International journal of molecular sciences, 19(8) (2018-08-01)
Obesity, characterized by excess lipid accumulation, has emerged as a leading public health problem. Excessive, adipocyte-induced lipid accumulation raises the risk of metabolic disorders. Adipose-derived stem cells (ASCs) are mesenchymal stem cells (MSCs) that can be obtained from abundant adipose
Young Taek Han et al.
Bioorganic & medicinal chemistry, 23(3), 579-587 (2014-12-24)
Through the fine tuning of the activity-sensitive aminoalkoxy moiety of 4,6-bisphenyl-2-(3-alkoxyanilino)pyrimidine as a novel inhibitor of the receptor for advanced glycation end products (RAGE), the tertiary amine was elucidated as an essential part associated with RAGE inhibition. On the basis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service