Skip to Content
Merck
All Photos(3)

Documents

536490

Sigma-Aldrich

2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine, polymer-bound

200-400 mesh, extent of labeling: 2.0-2.5 mmol/g loading, 1 % cross-linked

Synonym(s):

BEMP resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22

crosslinking

1 % cross-linked

reaction suitability

reaction type: solution phase peptide synthesis
reactivity: proton reactive

extent of labeling

2.0-2.5 mmol/g loading

particle size

200-400 mesh

SMILES string

C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.CCN(CC)P3(=NC(C)(C)C)N(C)CCCN3Cc4ccccc4

Looking for similar products? Visit Product Comparison Guide

Application

Polymer-supported base used for deprotonation and N-alkylation of weakly acidic heterocycles.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ying Wang et al.
Organic letters, 7(5), 925-928 (2005-02-25)
1,2,4-Oxadiazoles can be rapidly and efficiently synthesized from a variety of readily available carboxylic acids and amidoximes using either method A or method B. The use of commercially available polymer-supported reagents combined with microwave heating resulted in high yields and
A study of polymer-supported bases for the solution phase synthesis of glycosyl trichloroacetimidates.
Jose Luis Chiara, Lourdes Encinas and Beatriz Diaz
Tetrahedron Letters, 46, 2445-2448 (2005)
A convenient 'catch and release' synthesis of fused 2-alkylthio-pyrimidinones mediated by polymer-bound BEMP.
Gregory L. Adams et al.
Tetrahedron Letters, 44, 5041-5045 (2003)
Serena Gabrielli et al.
Molecules (Basel, Switzerland), 21(6) (2016-06-18)
Quinoline-2-carboxylates are an important subclass of quinoline derivatives largely present in a variety of biologically active molecules, as well as useful ligands in metal-catalyzed reactions. Herein, we present a new one-pot protocol for synthesizing this class of derivatives starting from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service