Skip to Content
Merck
All Photos(1)

Documents

79417

Sigma-Aldrich

Phosphazene base P2-Et

≥98.0% (NT)

Synonym(s):

1-Ethyl-2,2,4,4,4-pentakis(dimethylamino)-2λ5,4λ5-catenadi(phosphazene), Tetramethyl(tris(dimethylamino)phosphoranylidene)phosphorictriamid-Et-imin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3P=NP(=NC2H5)[N(CH3)2]2
CAS Number:
Molecular Weight:
339.40
Beilstein:
7485991
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (NT)

form

liquid

refractive index

n20/D 1.492 (lit.)
n20/D 1.492

bp

96 °C/0.05 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

SMILES string

CCN=P(N=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C12H35N7P2/c1-12-13-20(15(2)3,16(4)5)14-21(17(6)7,18(8)9)19(10)11/h12H2,1-11H3

InChI key

CFUKEHPEQCSIOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Phosphazene base P2-Et is an extremely strong, neutral nitrogen base. It gives excellent results where other bases fail

This strong non-ionic base has the ability to catalyze a wide variety of organic transformations such as:
  • Palladium-catalyzed cross-coupling reactions when used in combination with tBuXPhos or tBuBrettPhos G3 precatalysts.
  • Deprotonation of ortho-halobenzyl sulfones to generate α-sulfonyl benzylic carbanions.
  • Conversion of vinyl sufone to allyl sulfone.
  • α-Alkylation of 2-phenyl-2-oxazoline-4-carbonylcamphorsultam in the presence of tetrabutylammonium bromide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.H. Kim et al.
Tetrahedron Letters, 36, 4537-4537 (1995)
P2Et Phosphazene: A Mild, Functional Group Tolerant Base for Soluble, Room Temperature Pd-Catalyzed C?N, C?O, and C?C Cross-Coupling Reactions.
Buitrago S A, et al.
Organic Letters, 17(13), 3370-3373 (2015)
P2-Et-mediated deprotonation of ortho-halobenzyl sulfones: synthetic applications as zwitterionic synthons.
Costa A, et al.
Synlett, 2001(12), 1881-1884 (2001)
Enantioselective synthetic method for α-alkylserine via phase-transfer catalytic alkylation of 2-phenyl-2-oxazoline-4-carbonylcamphorsultam.
Lee J, et al.
The Journal of Organic Chemistry, 70(10), 4158-4161 (2005)
Phosphazene base P2-Et mediated isomerization of vinyl sulfones to allyl sulfones.
Jin Z, et al.
Tetrahedron Letters, 37(30), 5247-5248 (1996)

Articles

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Phosphazene base reagents are available as monomeric (P1 and BEMP), dimeric (P2), and tetrameric (P4) bases with different side chains to control their sterical hindrance.

Related Content

The Nicewicz lab is focused on the discovery of new and powerful reaction methodologies that proceed via the intermediacy of highly reactive cation radical species. Included in these transformations are anti-Markovnikov selective additions of amines, alcohols, carboxylic acids, amides and mineral acids to alkenes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service