Skip to Content
Merck
All Photos(1)

Key Documents

534757

Sigma-Aldrich

2-(4-Chlorophenyl)oxirane

96%

Synonym(s):

4-Chlorostyrene oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO
CAS Number:
Molecular Weight:
154.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5520 (lit.)

density

1.283 g/mL at 25 °C (lit.)

SMILES string

Clc1ccc(cc1)C2CO2

InChI

1S/C8H7ClO/c9-7-3-1-6(2-4-7)8-5-10-8/h1-4,8H,5H2

InChI key

IBWLXNDOMYKTAD-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

James W Comerford et al.
Molecules (Basel, Switzerland), 24(2) (2019-01-16)
Seaweed powder has been found to act as an effective catalyst for the fixation of CO₂ into epoxides to generate cyclic carbonates under solvent free conditions. Model background reactions were performed using metal halides and amino acids typically found in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service