Skip to Content
Merck
All Photos(1)

Key Documents

527858

Sigma-Aldrich

Aldolase Antibody 84G3, murine catalytic monoclonal antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.22

antibody product type

primary antibodies

storage temp.

−20°C

target post-translational modification

unmodified

Application

Novel biocatalyst for aldol condensation, especially for the re-facial addition of aliphatic ketones to aldehydes. Antibody 84G3 catalyzes aldol, crossed aldol, retroaldol, decarboxylation of β-keto acids, self aldol, and Robinson annulations.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong et al.
Angewandte Chemie (International ed. in English), 38(24), 3738-3741 (2000-01-29)
Nine efficient aldolase antibodies were generated by using hapten 1. This hapten unites reactive immunization and the transition state analogue approach in a single molecule. Characterization of two of these antibodies reveals that they are highly proficient (up to 1000-fold
S C Sinha et al.
Organic letters, 1(10), 1623-1626 (2000-06-03)
[formula: see text] Three monoclonal aldolase antibodies, generated against a beta-diketone hapten by reactive immunization, catalyzed rapid and highly enantioselective retro-aldol reactions of ent-8a-k, providing optically pure 8a-k by kinetic resolution. Compounds (+/-)-8a, (+/-)-8g, and (+/-)-8k have been resolved in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service