Recommended Products
Quality Level
Assay
90%
refractive index
n20/D 1.633 (lit.)
bp
141-143 °C/13 mmHg (lit.)
density
1.18 g/mL at 25 °C (lit.)
SMILES string
[H]C(=O)c1ccccc1SC
InChI
1S/C8H8OS/c1-10-8-5-3-2-4-7(8)6-9/h2-6H,1H3
InChI key
XIOBUABQJIVPCQ-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Rhodium-catalyzed synthesis of amides from aldehydes and azides by chelation-assisted C-H bond activation.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(32), 10511-10515 (2013-07-03)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service