Skip to Content
Merck
All Photos(4)

Key Documents

A9628

Sigma-Aldrich

2-Aminobenzaldehyde

≥98%

Synonym(s):

2-Formylaniline, Anthranilaldehyde, o-Aminobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

shipped in

dry ice

storage temp.

−20°C

SMILES string

Nc1ccccc1C=O

InChI

1S/C7H7NO/c8-7-4-2-1-3-6(7)5-9/h1-5H,8H2

InChI key

FXWFZIRWWNPPOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
  • Preparation of quinoline derivatives as antiviral agents
  • Preparation of electroluminescent materials for OLEDs
  • Friedlander-type synthesis
  • Preparation of 2-tosylaminophenyl cyclopropylmethanols for gold-catalyzed cyclopropyl carbinol rearrangement
  • Benzyl C-H bond amination of arylmethylamines catalyzed by hydroxy-TEMPO
  • Silver-catalyzed aniline mediated cascade hydroamination/cycloaddition reactions

Caution

Polymerizes rapidly at room temperature. May yield slightly hazy solution in ethanol due to the presence of a small amount of polymer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitin T Patil et al.
The Journal of organic chemistry, 77(14), 6179-6185 (2012-07-12)
DFT computational studies in the cyclization of aminoalkyne (see structure), which is generated in situ by 2-aminobenzaldehydes and terminal alkynes in the presence of metals and secondary amines, has been investigated. The study revealed that the mode of cyclization (exo
Hsien-Po Chiu et al.
Chembiochem : a European journal of chemical biology, 13(3), 364-366 (2012-01-10)
Sticky residue: Pyrroline-carboxy-lysine (Pcl) can be readily incorporated into proteins expressed in E. coli and mammalian cells by using the pyrrolysyl tRNA/tRNA synthetase pair. Pcl can be used as a single amino acid purification tag and can be site-specifically modified
Voula Mitsakos et al.
Bioorganic & medicinal chemistry, 19(4), 1535-1540 (2011-01-27)
The enzyme dihydrodipicolinate synthase (DHDPS) has been widely investigated as a target for new antibiotics. The o-aminobenzaldehyde (o-ABA) assay is routinely used as a highly specific, if qualitative, tool for DHDPS purification, whereby fractions containing active DHDPS appear purple upon
P Spiteller et al.
Phytochemistry, 57(3), 361-363 (2001-06-08)
Feeding experiments with 13C- and fluorine-labelled precursors were performed to reveal the biosynthesis of 2-aminobenzaldehyde in flowers of Robinia pseudoacacia and Philadelphus coronarius. The results are in agreement with the transformation of anthranilic acid to indole followed by oxidative ring
Yi-Feng Wang et al.
Chemistry, an Asian journal, 4(12), 1834-1838 (2009-10-16)
Generally, amine-catalyzed enantioselective transformations rely on chiral enamine or unsaturated iminium intermediates. Herein, we report a protocol involving dual activation by an aromatic iminium and hydrogen-bonding. An enantioselective aza-Michael-Henry domino reaction of 2-aminobenzaldehydes with nitroolefins has been developed through this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service