Skip to Content
Merck
All Photos(2)

Key Documents

516910

Sigma-Aldrich

2-Chloro-5-(chloromethyl)pyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5Cl2N
CAS Number:
Molecular Weight:
162.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

37-42 °C (lit.)

SMILES string

ClCc1ccc(Cl)nc1

InChI

1S/C6H5Cl2N/c7-3-5-1-2-6(8)9-4-5/h1-2,4H,3H2

InChI key

SKCNYHLTRZIINA-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Latli et al.
Journal of medicinal chemistry, 42(12), 2227-2234 (1999-06-23)
1-[(6-Chloro-3-pyridinyl)methyl]-2-imidazolidine (1), the N-desnitro metabolite of the major insecticide imidacloprid, is known to have similar potency to that of (-)-nicotine as an inhibitor of [3H](-)-nicotine binding at the rat recombinant alpha4beta2 neuronal nicotinic acetylcholine receptor (nAChR); IC50 values in the
2-Chloro-5-(chloromethyl) pyridine.
Feng ZQ, et al.
Acta Crystallographica Section E, Structure Reports Online, 67(2), o366-o366 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service