Skip to Content
Merck
All Photos(2)

Key Documents

485411

Sigma-Aldrich

Ammonium-14N2 sulfate

99.99 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(14NH4)2SO4
CAS Number:
Molecular Weight:
132.13
EC Number:
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:
NACRES:
NA.12

description

15N depleted

isotopic purity

99.99 atom % 14N

form

solid

mp

>280 °C (dec.) (lit.)

application(s)

agriculture

mass shift

depleted

SMILES string

[14NH3].[14NH3].OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)/i2*1+0;

InChI key

BFNBIHQBYMNNAN-ONPSQTMSSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazunori Sugiura et al.
Biochemical and biophysical research communications, 457(3), 242-248 (2015-01-17)
Intracellular redox state is a critical factor for fundamental cellular functions, including regulation of the activities of various metabolic enzymes as well as ROS production and elimination. Genetically-encoded fluorescent redox sensors, such as roGFP (Hanson, G. T., et al. (2004))
James F Hunter et al.
Environmental science & technology, 48(17), 10227-10234 (2014-08-06)
A large number of organic species emitted into the atmosphere contain cycloalkyl groups. While cyclic species are believed to be important secondary organic aerosol (SOA) precursors, the specific role of cyclic moieties (particularly for species with multiple or fused rings)
Christian Molitor et al.
Planta, 242(3), 519-537 (2015-02-24)
Aurone synthase belongs to the novel group 2 polyphenol oxidases and the presented kinetic characterization suggests a differing aurone biosynthesis in Asteraceae species compared to snapdragon. Aurone synthases (AUS) are polyphenol oxidases (PPO) physiologically involved in the formation of yellow
Yohei Y Yamamoto et al.
Journal of molecular biology, 426(14), 2667-2678 (2014-05-27)
Chaperonins are ubiquitous molecular chaperones with the subunit molecular mass of 60kDa. They exist as double-ring oligomers with central cavities. An ATP-dependent conformational change of the cavity induces the folding of an unfolded protein that is captured in the cavity.
Mostafizur Mazumder et al.
Nucleic acids research, 42(14), 9514-9522 (2014-07-25)
Modularity in controlling gene expression artificially is becoming an essential aspect of synthetic biology. Artificial transcriptional control of gene expression is one of the most well-developed methods for the design of novel synthetic regulatory networks. Such networks are intended to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service