Skip to Content
Merck
All Photos(1)

Key Documents

236233

Sigma-Aldrich

N-Methylaniline

≥99%

Synonym(s):

Monomethylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHCH3
CAS Number:
Molecular Weight:
107.15
Beilstein:
741982
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥99%

form

liquid

refractive index

n20/D 1.571 (lit.)

bp

196 °C (lit.)

mp

−57 °C (lit.)

solubility

water: slightly soluble 30 g/L
alcohol: soluble(lit.)
diethyl ether: soluble(lit.)

density

0.989 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CNc1ccccc1

InChI

1S/C7H9N/c1-8-7-5-3-2-4-6-7/h2-6,8H,1H3

InChI key

AFBPFSWMIHJQDM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical oxidation of N-methylaniline with dichromate (oxidant) has been studied by Raman spectroscopy.

Application

N-Methylaniline (NMA) was used in the preparation of self-assembled poly(N-methylaniline)-lignosulfonate (PNMA-LS) composite spheres with reactive silver-ion adsorbability. NMA was also used in electrodeposition of poly(N-methylaniline) (PNMA) coatings on a steel disc electrode using potentiodynamic, potentiostatic and galvanostatic techniques.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - STOT RE 2 Oral

Target Organs

Liver,spleen,Bone marrow

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electropolymerization of poly (N-methylaniline) on mild steel: Synthesis, characterization and corrosion protection.
Yag?an A, et al.
Journal of Electroanalytical Chemistry, 578(2), 231-238 (2005)
Qiu-Feng Lü et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(33), 10935-10944 (2013-07-03)
Self-assembled poly(N-methylaniline)-lignosulfonate (PNMA-LS) composite spheres with reactive silver-ion adsorbability were prepared from N-methylaniline by using lignosulfonate (LS) as a dispersant. The results show that the PNMA-LS composite consisted of spheres with good size distribution and an average diameter of 1.03-1.27
S Ramalingam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 76(1), 84-92 (2010-03-26)
The FTIR and FTRaman spectra of 2-bromo-4-methyl aniline (2-B-4-MA) molecule have been recorded using Brucker IFS 66V spectrometer in the range of 4000-100 cm(-1). The molecular geometry and vibrational frequencies in the ground state are calculated using the Hartree-Fock (HF)
Jonathan L Sessler et al.
Chemical communications (Cambridge, England), (14)(14), 1892-1894 (2005-03-30)
A novel porphyrin-fullerene dyad assembled through Watson-Crick hydrogen bonds is described; this system undergoes photoinduced electron transfer upon irradiation with visible light to produce a charge separated state that is substantially longer lived than that of previous dyads of this
Israel González et al.
Organic letters, 11(8), 1677-1680 (2009-04-10)
N-Methylanilines are readily synthesized in high yields through the copper(II)-promoted coupling of anilines and methylboronic acid. This method represents a new approach for the selective monomethylation of anilines, and it is the first reported example of a Chan-Lam coupling involving

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service