Skip to Content
Merck
All Photos(1)

Documents

49605

Sigma-Aldrich

L-Glutamic acid hemimagnesium salt tetrahydrate

≥98.0% (NT)

Synonym(s):

Magnesium L-glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16MgN2O8 · 4H2O
CAS Number:
Molecular Weight:
388.61
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (NT)

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

application(s)

peptide synthesis

SMILES string

O.O.O.O.N[C@@H](CCC(=O)O[Mg]OC(=O)CC[C@H](N)C(O)=O)C(O)=O

InChI

1S/2C5H9NO4.Mg.4H2O/c2*6-3(5(9)10)1-2-4(7)8;;;;;/h2*3H,1-2,6H2,(H,7,8)(H,9,10);;4*1H2/q;;+2;;;;/p-2/t2*3-;;;;;/m00...../s1

InChI key

ZWCGDRSVJYJGFY-GYDBBPQESA-L

General description

L-Glutamic acid hemimagnesium salt tetrahydrate is commonly known as Magnesium L-glutamate. It donates magnesium ions necessary for effective protein synthesis.

Application

L-Glutamic acid hemimagnesium salt tetrahydrate is a magnesium transfer agent/carrier. It can also be used in the preparation of S30A buffer solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshiki Akui et al.
iScience, 24(8), 102859-102859 (2021-08-14)
Biochemical systems in living cells have their optimum concentration ratio among each constituent element to maintain their functionality. However, in the case of the biochemical system with complex interactions and feedbacks among elements, their activity as a system greatly changes
Efficient incorporation of unnatural amino acids into proteins with a robust cell-free system
W Gao
Methods and Protocols, 2, 16-16 (2019)
Simplifying and streamlining Escherichia coli-based cell-free protein synthesis
WC Yang
Biotechnology Progress, 28, 413-420 (2012)
Synthesis of 2.3 mg/ml of protein with an all Escherichia coli cell-free transcription?translation system.
Caschera F and Noireaux V
Biochimie, 99, 162-168 (2014)
Synthesis and antiarrhythmic activity of new biologically active potassium and magnesium donors.
Berlyand A S, et al.
Pharmaceutical Chemistry Journal, 41(12), 634-637 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service