Skip to Content
Merck
All Photos(1)

Key Documents

49621

Sigma-Aldrich

L-Glutamic acid monosodium salt monohydrate

≥98.0% (NT), for peptide synthesis

Synonym(s):

L-2-Aminopentanedioic acid, MSG, Sodium L-glutamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaOOCCH2CH2CH(NH2)COOH · H2O
CAS Number:
Molecular Weight:
187.13
Beilstein:
4164348
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Product Name

L-Glutamic acid monosodium salt monohydrate, ≥98.0% (NT)

Quality Level

Assay

≥98.0% (NT)

form

solid

optical activity

[α]20/D −4.2±0.3°, c = 1.2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

232 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O

InChI

1S/C5H9NO4.Na.H2O/c6-3(5(9)10)1-2-4(7)8;;/h3H,1-2,6H2,(H,7,8)(H,9,10);;1H2/q;+1;/p-1/t3-;;/m0../s1

InChI key

GJBHGUUFMNITCI-QTNFYWBSSA-M

Looking for similar products? Visit Product Comparison Guide

General description

L-Glutamic acid monosodium salt monohydrate, commonly known as monosodium glutamate (MSG), is a sodium salt of glutamic acid.

Application

L-Glutamic acid monosodium salt monohydrate is used in carboxylation reactions and acts as a precursor for the synthesis and precipitation of α L-glutamic acid

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Redox hydrogel-based bienzyme microelectrodes for amperometric monitoring of L-glutamate.
Mikeladze E, et al.
Electroanalysis, 14(6), 393-393 (2002)
Influence of pH Value and Ionic Liquids on the Solubility of l-Alanine and l-Glutamic Acid in Aqueous Solutions at 30? C
M Voges, et al.
Journal of Chemical and Engineering Data, 62, 52-61 (2017)
Precipitation of ? L-glutamic acid: determination of growth kinetics
J Scholl, et al.
Faraday Discussions, 136, 247-264 (2007)
Inclusion of the stable form of a polymorph within crystals of its metastable form.
Cashell C, et al.
Crystal Growth & Design, 3(6), 869-872 (2003)
Spontaneous exfoliation of large-sized graphene oxide with low defect concentration by simple wet chemistry
WJ Lee, et al.
Carbon, 182, 214-222 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service