Skip to Content
Merck
All Photos(1)

Key Documents

49360

Sigma-Aldrich

Glutaconic acid

97% (T)

Synonym(s):

2-Pentenedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH=CHCOOH
CAS Number:
Molecular Weight:
130.10
Beilstein:
1759560
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97% (T)

functional group

carboxylic acid

SMILES string

OC(=O)C\C=C\C(O)=O

InChI

1S/C5H6O4/c6-4(7)2-1-3-5(8)9/h1-2H,3H2,(H,6,7)(H,8,9)/b2-1+

InChI key

XVOUMQNXTGKGMA-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Glutaconic acid, also known as 2-pentenedioic acid, is an unsaturated dicarboxylic acid. It is formed as one of the degradation products during the partial wet oxidation (PWO) of alkali lignin. The analysis of its crystal structure indicates that the compound exists predominantly in the trans-conformation. The geometric bond lengths and bond angles of glutaconic acid have been obtained using Hartree–Fock (HF), density functional calculations and IR spectral data.

Application

Glutaconic acid has been used in the preparation of CoA-substrate glutaconyl-CoA by reacting with acetyl-CoA. It may be used in the preparation of 6-chloro-2(2H)-pyranone by reacting with phosphorus pentachloride (PCl5).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Crystal and molecular structure of glutaconic acid.
Thomas L and Srikrishnan T.
Journal of Chemical Crystallography, 33(9), 689-693 (2003)
Theoretical studies of molecular structure and vibrational spectra of glutaconic acid.
Atalay Y, et al.
Journal of Molecular Structure, 787(1), 90-95 (2006)
H Osaka et al.
Brain & development, 15(2), 125-127 (1993-03-01)
A 10-month-old male with glutaric aciduria type-1 (GA-1) is reported. This patient showed frequent partial motor seizures, irritability, and involuntary movements, including oral dyskinesia at the age of 3 months. On admission, magnetic resonance (MR) scanning revealed a chronic subdural
6-Chloro-2(2H)-pyranone: a new 2(2H)-pyranone synthon.
Biagetti M, et al.
Tetrahedron Letters, 44(3), 607-610 (2003)
Alkaline Partial Wet Oxidation of Lignin for the Production of Carboxylic Acids.
Demesa AG, et al.
Chemical Engineering & Technology, 38(12), 2270-2278 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service