Skip to Content
Merck
All Photos(1)

Key Documents

493066

Sigma-Aldrich

N,N-Dimethylethanolamine-15N

98 atom % 15N

Synonym(s):

N,N-Dimethylaminoethanol-15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH215N(CH3)2
CAS Number:
Molecular Weight:
90.13
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % 15N

Quality Level

refractive index

n20/D 1.429 (lit.)

bp

133-134 °C (lit.)

mp

−70 °C (lit.)

density

0.897 g/mL at 25 °C

mass shift

M+1

SMILES string

C[15N](C)CCO

InChI

1S/C4H11NO/c1-5(2)3-4-6/h6H,3-4H2,1-2H3/i5+1

InChI key

UEEJHVSXFDXPFK-HOSYLAQJSA-N

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

102.2 °F - closed cup

Flash Point(C)

39.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service