Skip to Content
Merck
All Photos(1)

Key Documents

48220

Sigma-Aldrich

Gadolinium(III) oxide

≥99.9%

Synonym(s):

Digadolinium trioxide, Gadolinia, Gadolinium oxide, Gadolinium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd2O3
CAS Number:
Molecular Weight:
362.50
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9%

reaction suitability

reagent type: catalyst
core: gadolinium

mp

2330 °C (lit.)

density

7.407 g/mL at 20 °C (lit.)

SMILES string

O=[Gd]O[Gd]=O

InChI

1S/2Gd.3O

InChI key

RNYFGGMJZRTZKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eva Hemmer et al.
Acta biomaterialia, 9(1), 4734-4743 (2012-09-12)
Bioimaging is an important diagnostic tool in the investigation and visualization of biological phenomena in cells and in medicine. In this context, up-converting Gd(2)O(3):Er(3+),Yb(3+) nanostructures (nanoparticles, nanorods) have been synthesized by precipitation methods and hydrothermal synthesis. Independent of size and
Yuanzhi Shao et al.
Biomaterials, 33(27), 6438-6446 (2012-06-19)
The feasibility of the gadolinium-doped mesoporous silica nanocomposite Gd(2)O(3)@MCM-41 as a safe, effective MRI nanoprobe has been validated in the current investigation systematically from atomistic and molecular modeling to its synthesis and characterization on in vivo MR imaging and biocompatibility.
Natalia Abrikossova et al.
Nanotechnology, 23(27), 275101-275101 (2012-06-19)
We have previously shown that gadolinium oxide (Gd(2)O(3)) nanoparticles are promising candidates to be used as contrast agents in magnetic resonance (MR) imaging applications. In this study, these nanoparticles were investigated in a cellular system, as possible probes for visualization
Taejong Paik et al.
ACS nano, 7(3), 2850-2859 (2013-02-26)
Here, we report the shape-controlled synthesis of tripodal and triangular gadolinium oxide (Gd2O3) nanoplates. In the presence of lithium ions, the shape of the nanocrystals is readily controlled by tailoring reaction parameters such as temperature and time. We observe that
Luc Faucher et al.
ACS applied materials & interfaces, 4(9), 4506-4515 (2012-07-28)
Ultrasmall paramagnetic Gd(2)O(3) nanoparticles have been developed as contrast agents for molecular and cellular preclinical MRI procedures. These small particles (mean diameter <5 nm) have the highest Gd density of all paramagnetic contrast agents. They generate strong positive contrast enhancement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service