Skip to Content
Merck
All Photos(2)

Key Documents

278513

Sigma-Aldrich

Gadolinium(III) oxide

powder, 99.9% trace metals basis

Synonym(s):

Digadolinium trioxide, Gadolinia, Gadolinium oxide, Gadolinium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Gd2O3
CAS Number:
Molecular Weight:
362.50
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: gadolinium

mp

2330 °C (lit.)

density

7.407 g/mL at 20 °C (lit.)

SMILES string

O=[Gd]O[Gd]=O

InChI

1S/2Gd.3O

InChI key

RNYFGGMJZRTZKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdiel Inggrid Setyawati et al.
Journal of biomedical materials research. Part A, 101(3), 633-640 (2012-08-29)
Engineered nanomaterials have become prevalent in our everyday life. While the popularity of using nanomaterials in consumer products continues to rise, increasing awareness of nanotoxicology has also fuelled efforts to accelerate our understanding of the ill effects that different nanomaterials
Taejong Paik et al.
ACS nano, 7(3), 2850-2859 (2013-02-26)
Here, we report the shape-controlled synthesis of tripodal and triangular gadolinium oxide (Gd2O3) nanoplates. In the presence of lithium ions, the shape of the nanocrystals is readily controlled by tailoring reaction parameters such as temperature and time. We observe that
Nikesh Gupta et al.
International journal of nanomedicine, 7, 5491-5500 (2012-12-13)
Mesoporous silica nanoparticles coencapsulating gadolinium oxide and horseradish peroxidase (HRP) have been synthesized in the aqueous core of sodium bis-(2-ethylhexyl) sulfosuccinate (AOT)-hexane-water reverse micelle. The average diameter of these silica particles is around 25 nm and the particles are spherical
Andrée-Anne Guay-Bégin et al.
Langmuir : the ACS journal of surfaces and colloids, 28(1), 774-782 (2011-10-06)
The performance of nanomaterials for biomedical applications is highly dependent on the nature and the quality of surface coatings. In particular, the development of functionalized nanoparticles for magnetic resonance imaging (MRI) requires the grafting of hydrophilic, nonimmunogenic, and biocompatible polymers
Weihua Di et al.
Biomaterials, 32(29), 7226-7233 (2011-07-13)
Multifunctional materials for biological use have mostly been designed with composite or hybrid nanostructures in which two or more components are incorporated. The present work reports on a multifunctional biomaterial based on single-phased luminescent mesoporous lanthanide oxide nanoparticles that combine

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service