Skip to Content
Merck
All Photos(1)

Key Documents

48190

Sigma-Aldrich

(±)-1-(2-Furyl)ethanol

≥99.0% (GC)

Synonym(s):

(±)-α-Methylfuran-2-methanol, (±)-2-Furyl methyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8O2
CAS Number:
Molecular Weight:
112.13
Beilstein:
107814
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

contains

~0.05% hydroquinone as stabilizer

refractive index

n20/D 1.479

bp

167-170 °C

density

1.078 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(O)c1ccco1

InChI

1S/C6H8O2/c1-5(7)6-3-2-4-8-6/h2-5,7H,1H3

InChI key

UABXUIWIFUZYQK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(±)-1-(2-Furyl)ethanol [(±)-2-Furyl methyl carbinol] is a furan derivative. The synthesis of 4-hydroxy-2-methylcyclopent-2-en-1-one from 2-furyl methyl carbinol has been reported.

Application

(±)-1-(2-Furyl)ethanol (Racemic 1-(2-furyl)ethanol) may be used in the synthesis of 1-acetoxy-1-[2-furyl]ethan.

Caution

may discolor to brown on storage

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

An Enantioselective Total Synthetic Approach to (+)-Heptemerone G and (+)-Guanacastepene A from 2-Furyl Methyl Carbinol.
Michalak K and Wicha J.
Synlett, 24, 1387-1390 (2013)
A Ghanem et al.
Chirality, 13(2), 118-123 (2001-02-15)
Asymmetric acetylation of racemic 1-(2-furyl)ethanol with the innocuous acyl donor isopropenyl acetate catalyzed by lipases in organic media afforded the chiral alcohol and acetate in high enantiomeric excess (up to 99%). The effect of molecular sieves as well as organic
Jordan Lopez et al.
Journal of agricultural and food chemistry, 67(41), 11444-11453 (2019-10-09)
Innovative approaches to develop flavors with high sensory appeal are critical in encouraging increased consumer preference and adoption of low sodium foods. Gas chromatography-olfactometry, coupled with stable isotope dilution assays and sensory experiments, led to the identification of the odorants
Zhen Zeng et al.
Food chemistry, 188, 591-595 (2015-06-05)
The formation of 2-vinylfuran from the corresponding 4-oxo-2-hexenal (OHE, a lipid oxidation product) under the catalysis of amino acid were studied. The effects of amino acids, reaction temperature, reaction time, water content, pH, metallic ions and some food additives on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service