Skip to Content
Merck
All Photos(2)

Key Documents

477222

Sigma-Aldrich

4-Chloroaniline

purified by sublimation, ≥99%

Synonym(s):

1-Amino-4-chlorobenzene, 4-Chlorophenylamine, p-Chloroaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NH2
CAS Number:
Molecular Weight:
127.57
Beilstein:
471359
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.4 (vs air)

Quality Level

vapor pressure

0.15 mmHg ( 25 °C)

Assay

≥99%

form

crystals
powder
solid

purified by

sublimation

bp

232 °C (lit.)

mp

67-70 °C (lit.)

functional group

chloro

SMILES string

Nc1ccc(Cl)cc1

InChI

1S/C6H6ClN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

QSNSCYSYFYORTR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Chloroaniline (4-ClA) is a chlorinated aromatic amine that is formed as an intermediate during the microbial decomposition of phenylurea and phenylcarbamate. The formation of various oligomers by polymerization of 4-ClA with guaiacol in an aqueous solution containing oxidoreductases has been reported. 4-ClA undergoes decomposition via oxidation by fusarium oxysporum, a soil fungus.

Application

4-Chloroaniline may be used to synthesize:
  • 4-chloronitrosobenzene
  • 2-bromo-4-chloroaniline
  • 2,6-dibromo-4-chloroaniline

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

248.0 °F - closed cup

Flash Point(C)

120.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative co-oligomerization of guaiacol and 4-chloroaniline.
Simmons KE, et al.
Environmental Science & Technology, 23(1), 115-121 (1989)
The action of chloride peroxidase on 4-chloroaniline. N-oxidation and ring halogenation.
Corbett MD, et al.
The Biochemical Journal, 187(3), 893-903 (1980)
M D Corbett et al.
The Biochemical journal, 175(2), 353-360 (1978-11-01)
The incubation of 4-chloroaniline with chloroperoxidase and H2O2 resulted in a rapid formation of 4-chloronitrosobenzene. This enzymic oxidation displayed a pH optimum at 4.4 with a Km of 8.1x10(-4)M and catalytic-centre activity of 312. The initial rate of the reaction
Microbial oxidation of 4-chloroaniline.
D D Kaufman et al.
Journal of agricultural and food chemistry, 21(1), 127-132 (1973-01-01)
D Mortenson et al.
International endodontic journal, 45(9), 878-882 (2012-04-11)
To determine if the formation of para-chloroaniline (PCA) can be avoided by using an alternative irrigant following sodium hypochlorite but before chlorhexidine. Fifty-five single-rooted teeth were decoronated, instrumented to size 40, .06 taper whilst being irrigated with 14% ethylene-diamine-tetra-acetic acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service