Skip to Content
Merck
All Photos(2)

Key Documents

466875

Sigma-Aldrich

Poly(ethylene glycol) (18) tridecyl ether

n=18, tridecyl ether group is a mixture of C11 to C14 iso-alkyl ethers with C13 iso-alkyl predominating

Synonym(s):

Polyoxyethylene (18) tridecyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C13H27(OCH2CH2)nOH, n~18
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

refractive index

n20/D 1.464 (lit.)

mp

28 °C (lit.)

hydroxyl value

54‑62 mg KOH/g

solubility

water, isopropanol: 1 wt. % at 25 °C

density

0.96 g/mL at 25 °C (lit.)

HLB

15

SMILES string

O(CCO)CCCCCCCCCCCCC

InChI

1S/C15H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-14-17-15-13-16/h16H,2-15H2,1H3

InChI key

OJDDJQYSKDIXOE-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Shiibashi et al.
Developmental and comparative immunology, 25(5-6), 461-465 (2001-05-18)
NADPH oxidase has been identified as the superoxide-generating enzyme in fish neutrophils. To clarify the electron-donating ability of this enzyme, we examine the requirement of NADPH as the electron donor in superoxide generation in tilapia (Oreochromis niloticus) neutrophils using CLA-dependent
R E Mayner et al.
Journal of biological standardization, 15(1), 1-10 (1987-01-01)
The glycoprotein content of rabies vaccines containing the Pitman-Moore strain of rabies virus was measured by the single radial immunodiffusion assay and correlated with vaccine potency. The variability of this assay was 6.3% for a single vaccine lot tested over
Pitfalls in the use of commercial nonionic detergents for the solubilization of integral membrane proteins: sulfhydryl oxidizing contaminants and their elimination.
H W Chang et al.
Analytical biochemistry, 104(1), 112-117 (1980-05-01)
H Suzuki et al.
Journal of biochemistry, 93(3), 709-715 (1983-03-01)
The relationship between glucose metabolism and the "respiratory burst" of phagocytosing polymorphonuclear leukocytes (PMN) was studied in a Renex 30-treated cell system of guinea pig PMN by a polarometric technique. Phagocytosing PMN were treated with a detergent (Renex 30) and
M I Aveldaño
Archives of biochemistry and biophysics, 324(2), 331-343 (1995-12-20)
The solubilization of rhodopsin and phospholipids from disks prepared from bovine retinal rods was studied using five different detergents. The relative amounts of rhodopsin and lipid extracted during membrane solubilization differed dramatically with the nature of the surfactant; the two

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service