Skip to Content
Merck
All Photos(1)

Key Documents

463760

Sigma-Aldrich

Ruthenium(IV) oxide hydrate

99.9% trace metals basis

Synonym(s):

Hydrous ruthenium oxide, Ruthenium dioxide hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
RuO2 · xH2O
CAS Number:
Molecular Weight:
133.07 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

SMILES string

O.O=[Ru]=O

InChI

1S/H2O.2O.Ru/h1H2;;;

InChI key

FGEKTVAHFDQHBU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aleksandar R Zeradjanin et al.
ChemSusChem, 5(10), 1897-1904 (2012-08-16)
The reaction path of the Cl(2) evolution reaction (CER) was investigated by combining electrochemical and spectroscopic methods. It is shown that oxidation and reconstruction of the catalyst surface during CER is a consequence of the interaction between RuO(2) and water.
Naiqiang Yan et al.
Environmental science & technology, 45(13), 5725-5730 (2011-06-15)
Catalytic conversion of elemental mercury (Hg(0)) to its oxidized form has been considered as an effective way to enhance mercury removal from coal-fired power plants. In order to make good use of the existing selective catalytic reduction of NO(x) (SCR)
O Delmer et al.
Physical chemistry chemical physics : PCCP, 11(30), 6424-6429 (2009-10-08)
The chemical potential of a component of a binary metastable compound is considered in the single-phase and in the compositionally non-variant two-phase regime. A detailed thermodynamic analysis reveals striking differences for identical nominal compositions. Without the loss of generality the
Hiroshi Nishiyama et al.
ChemSusChem, 4(2), 208-215 (2011-02-18)
Sodium, niobium, and tantalum phosphate bronzes Na(4)M(8)P(4)O(32) (M=Nb, Ta) are employed as photocatalysts for water splitting to reveal the effects of the distortion of metal-oxygen octahedra on the photocatalytic performance. Addition of RuO(2) as a co-catalyst leads to high, stable
Chengzhi Hu et al.
Journal of environmental sciences (China), 24(2), 297-302 (2012-06-05)
A polyaluminum containing a high concentration of Al13 polymer and active chlorine (PACC) was successfully synthesized by a new electrochemical reactor using Ti/RuO2-TiO2 anodes. PACC can potentially be used as a dual-function chemical reagent for water treatment. The obtained results

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service