Skip to Content
Merck
All Photos(6)

Key Documents

P23954

Sigma-Aldrich

m-Phenylenediamine

flakes, 99%

Synonym(s):

1,3-Benzenediamine, 1,3-Diaminobenzene, 1,3-Phenylenediamine, MPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(NH2)2
CAS Number:
Molecular Weight:
108.14
Beilstein:
471357
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

3.7 (vs air)

Quality Level

vapor pressure

0.62 mmHg ( 100 °C)

Assay

99%

form

flakes

autoignition temp.

1040 °F

bp

282-284 °C

mp

64-66 °C

SMILES string

Nc1cccc(N)c1

InChI

1S/C6H8N2/c7-5-2-1-3-6(8)4-5/h1-4H,7-8H2

InChI key

WZCQRUWWHSTZEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

m-Phenylenediamine (MPD) is an aromatic diamine. It copolymerizes with iso- or terephthaloyl chloride and 3,5-diaminobenzoic acid or 2,4-diaminobenzene sulfonic acid (or sulfonate) to afford copolyamides by low temperature solution polymerization. MPD has been reported to be formed selectively during the hydrogenation of m-dinitrobenzene in the presence of bimetallic Ni-Pt catalysts supported on carbon. Cyclic voltammetric method has been proposed to study the electrochemical copolymerization of aniline with MPD in sulfuric acid solution.

Application

m-Phenylenediamine may be used in the synthesis of the following:
  • intrinsically electrically semiconducting microparticles of semiladder poly(m-phenylenediamine-co-2-hydroxy-5-sulfonic aniline) structures
  • extraction medium based on chitosan-poly(m-phenylenediamine) (CS-PPD) Fe3O4 nanocomposite, used as sorbent for the magnetic solid-phase extraction (MSPE) of polychlorinated biphenyls
  • series of terpolymers, via chemical oxidative polymerization
  • thin film composite (TFC) membranes based polyamide
  • TFC reverse osmosis (RO) membranes

Other Notes

May darken in storage

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qie Gen Liao et al.
Analytical and bioanalytical chemistry, 406(29), 7571-7579 (2014-10-25)
A extraction medium based on chitosan-poly(m-phenylenediamine) (CS-PPD) @Fe3O4 nanocomposite was synthesized by chemical polymerization of m-phenylenediamine in the presence of chitosan coated magnetic nanocomposite, and for the first time, used as the sorbent for the magnetic solid-phase extraction (MSPE) of
Electrochemical copolymerization of aniline with m-phenylenediamine.
Mazeikiene R and Malinauskas A.
Synthetic Metals, 92(3), 259-263 (1998)
Sudakar Padmanaban et al.
Scientific reports, 7, 44796-44796 (2017-03-21)
Silver nanobelts (AgNBs) have attracted a great interest due to their excellent electrical conductivity and mechanical strength, leading a facile synthesis of these AgNBs in great demand. In here, we are reporting a simple, aqueous phase, size tunable synthesis of
Polyamide thin film composite membrane prepared from m-phenylenediamine and m-phenylenediamine-5-sulfonic acid.
Yong Z, et al.
Journal of Membrane Science, 270(1), 162-168 (2006)
Preparation and characterization of soluble terpolymers from m-phenylenediamine, o-anisidine, and 2, 3-xylidine.
Li X-G, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 39(22), 3989-4000 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service