Skip to Content
Merck
All Photos(2)

Key Documents

460036

Sigma-Aldrich

5-Norbornene-2-endo,3-exo-dicarboxylic acid

97%

Synonym(s):

NDC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O4
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

mp

185-189 °C (lit.)

SMILES string

OC(=O)[C@@H]1[C@@H]2C[C@@H](C=C2)[C@H]1C(O)=O

InChI

1S/C9H10O4/c10-8(11)6-4-1-2-5(3-4)7(6)9(12)13/h1-2,4-7H,3H2,(H,10,11)(H,12,13)/t4-,5+,6-,7-/m1/s1

InChI key

NIDNOXCRFUCAKQ-XZBKPIIZSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellis J Powell et al.
Laboratory animals, 52(4), 402-412 (2018-01-13)
Severe combined immunodeficiency (SCID) is defined by the lack of an adaptive immune system. Mutations causing SCID are found naturally in humans, mice, horses, dogs, and recently in pigs, with the serendipitous discovery of the Iowa State University SCID pigs.
Sofia R Gameiro et al.
Oncotarget, 5(2), 403-416 (2014-02-01)
Radiation therapy (RT) is used for local tumor control through direct killing of tumor cells. Radiation-induced cell death can trigger tumor antigen-specific immune responses, but these are often noncurative. Radiation has been demonstrated to induce immunogenic modulation (IM) in various
Christiana J Johnson et al.
Molecular vision, 14, 2211-2226 (2008-12-06)
Our goal was to improve and standardize the procedure for subretinal injection of mouse eyes. Also, we wished to optimize conditions for electroporation of retinal pigment epithelium (RPE) cells in the mouse eye with naked plasmids. Mouse eyes were injected
Xuerong Wen et al.
Neurology, 84(9), 944-950 (2015-02-06)
The study aims were to investigate secular trends in antiepileptic drug (AED) use in women during pregnancy, and to compare the use of first- and second-generation AEDs. Study participants consisted of female Florida Medicaid beneficiaries, older than 15 years, and
Omer Raheem et al.
Journal of translational medicine, 9, 185-185 (2011-11-01)
Prostate cancer metastasizes to bone in the majority of patients with advanced disease leading to painfully debilitating fractures, spinal compression and rapid decline. In addition, prostate cancer bone metastases often become resistant to standard therapies including androgen deprivation, radiation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service