Skip to Content
Merck
All Photos(1)

Key Documents

456500

Sigma-Aldrich

Methyl 4-methoxybutyrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2)3CO2CH3
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
98%

Quality Level

Assay

98%

refractive index

n20/D 1.408 (lit.)

bp

162-164 °C/767 mmHg (lit.)

density

0.969 g/mL at 25 °C (lit.)

functional group

ester
ether

SMILES string

COCCCC(=O)OC

InChI

1S/C6H12O3/c1-8-5-3-4-6(7)9-2/h3-5H2,1-2H3

InChI key

VHDGWXQBVWAMJA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

129.2 °F - closed cup

Flash Point(C)

54 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Alemán et al.
The Journal of organic chemistry, 70(19), 7731-7736 (2005-09-10)
[structure: see text] A systematic conformational study of omega-methoxy methyl esters, CH(3)O-(CH2)n-COO-CH3 with n = 3 and 4, has been performed using quantum mechanical calculations at the MP2 level. Calculations have been carried out in both gas phase and chloroform
Maria Cammenberg et al.
Chembiochem : a European journal of chemical biology, 7(11), 1745-1749 (2006-09-23)
One of the commercial methods for preparing enantiopure amines is lipase-catalyzed kinetic resolution, although lipases catalyze aminolysis with only low activity. Interestingly, in 1997 Balkenhohl et al. used ethyl methoxyacetate instead of ethyl butyrate as an acylation reagent for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service