Skip to Content
Merck
All Photos(1)

Documents

281026

Sigma-Aldrich

Tetrabutylammonium hexafluorophosphate

98%

Synonym(s):

TBAPF6, Tetra-n-butylammonium hexafluorophosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(PF6)
CAS Number:
Molecular Weight:
387.43
Beilstein:
4056584
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

244-246 °C (lit.)

SMILES string

F[P-](F)(F)(F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.F6P/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-7(2,3,4,5)6/h5-16H2,1-4H3;/q+1;-1

InChI key

BKBKEFQIOUYLBC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium hexafluorophosphate (TBAPF6) can be used:
  • As a catalyst in the synthesis of 3,4-dihydropyrimidin-2(1H)-one and 3,4-dihydropyrimidine-2(1H)-thione derivatives via Biginelli reaction.
  • Along with 1-ethyl-3-methyl imidazolium hexafluorophosphate as a supporting electrolyte in non-aqueous redox flow batteries.
  • As a supporting electrolyte in the electropolymerisation.

Tetrabutylammonium hexafluorophosphate can be used as a supporting electrolyte for the formation of cyclic disulfide moiety of 3,5-diimido-1,2-dithiolane derivatives by S-S coupling of dithioanilides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical synthesis and properties of copolymers based on N-alkylpyrroles and thiophene
Vautrin M, et al.
Electrochemical Communications, 1(6), 233-237 (1999)
Holger Hartleb et al.
ACS nano, 9(10), 10461-10470 (2015-09-19)
We have investigated the photophysical properties of electrochemically gate-doped semiconducting single-wall carbon nanotubes (s-SWNTs). A comparison of photoluminescence (PL) and simultaneously recorded absorption spectra reveals that free-carrier densities correlate well with the first sub-band exciton or trion oscillator strengths but
Tetrabutylammonium hexafluorophosphate and 1-ethyl-3-methyl imidazolium hexafluorophosphate ionic liquids as supporting electrolytes for non-aqueous vanadium redox flow batteries
Zhang D, et al.
Journal of Power Sources, 203, 201-205 (2012)
Electrochemical Formation of 3, 5-Diimido-1, 2-dithiolanes by Dehydrogenative Coupling
Breising VM, et al.
Organic Letters (2018)
The application of a non-aqueous bis (acetylacetone) ethylenediamine cobalt electrolyte in redox flow battery
Zhang D, et al.
Journal of Power Sources, 217, 199-203 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service