Skip to Content
Merck
All Photos(1)

Documents

452505

Sigma-Aldrich

2,6-Di(tert-butyl-d9)-4-methyl(phenol-3,5,O-d3)

≥98 atom % D, ≥98% (CP)

Synonym(s):

BHT-d21

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CD3)3C]2C6D2(CH3)OD
CAS Number:
Molecular Weight:
241.48
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥98% (CP)

form

solid

bp

265 °C (lit.)

mp

69-71 °C (lit.)

mass shift

M+21

SMILES string

[2H]Oc1c(c([2H])c(C)c([2H])c1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H])C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3/i2D3,3D3,4D3,5D3,6D3,7D3,8D,9D/hD

InChI key

NLZUEZXRPGMBCV-YLENQQAOSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruirui Zhang et al.
Journal of agricultural and food chemistry, 68(16), 4670-4677 (2020-02-18)
One typical synthetic phenolic antioxidant 2,6-di-tert-butyl-hydroxytoluene (BHT) is widely used in foodstuff. Concerns are rising on the toxicity of BHT and its metabolites through dietary exposure. In this study, the effects of food macronutrients (i.e., lipid, carbohydrate, fiber, protein, and
Wei Wang et al.
Environmental science & technology, 50(1), 428-434 (2015-12-03)
Synthetic phenolic antioxidants (SPAs), including 2,6-di-tert-butyl-4-hydroxytoluene (BHT), are extensively used in food, cosmetic and plastic industries. Nevertheless, limited information is available on human exposures, other than the dietary sources, to SPAs. In this study, occurrence of 9 SPAs and their
Runzeng Liu et al.
Environmental pollution (Barking, Essex : 1987), 221, 227-233 (2016-12-06)
In this study, seven synthetic phenolic antioxidant (SPA) analogues were positively found in urban and rural indoor dust samples collected from Shandong province in China, among which the novel 2,4,6-tri-tert-butylphenol (AO 246), 2,6-di-tert-butyl-4-sec-butylphenol (DTBSBP), 2,4-di-tert-butylphenol (DBP) and 4,4'-butylidenebis (2-(1,1-dimethylethyl)-5- methyl-phenol)
Wei Wang et al.
Environment international, 128, 24-29 (2019-04-29)
Synthetic phenolic antioxidants (SPAs) such as 2,6-di-tert-butyl-4-hydroxytoluene (butylated hydroxytoluene, BHT), are used in a wide variety of consumer products, including certain foodstuffs (e.g. fats and oils) and cosmetics. Although BHT is considered generally safe as a food preservative when used
Lucia Gusmaroli et al.
Analytical and bioanalytical chemistry, 410(17), 4165-4176 (2018-04-25)
During the last decades, the quality of aquatic ecosystems has been threatened by increasing levels of pollutions, caused by the discharge of man-made chemicals, both via accidental release of pollutants as well as a consequence of the constant outflow of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service