Skip to Content
Merck
All Photos(1)

Key Documents

B1215000

Butylhydroxytoluene

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

autoignition temp.

878 °F

manufacturer/tradename

EDQM

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical

format

neat

storage temp.

2-8°C

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mateusz Maciejczyk et al.
Nutrients, 10(10) (2018-10-20)
A high-sucrose diet (HSD) is widely known for its cariogenic effects and promotion of obesity, insulin resistance, type 2 diabetes, and cancer. However, the impact of the HSD diet on the salivary gland function as well as the level of
Hiroaki Gotoh et al.
Science advances, 4(10), eaat7629-eaat7629 (2018-10-20)
An elastomer is a three-dimensional network with a cross-linked polymer chain that undergoes large deformation with a small external force and returns to its original state when the external force is removed. Because of this hyperelasticity, elastomers are regarded as
Adam Dąbrowski et al.
Antioxidants (Basel, Switzerland), 9(2) (2020-02-27)
Oxidative stress underlies the pathomechanisms of toxic action of cadmium (Cd), including its damaging impact on the oral cavity. This study investigated whether the administration of an extract from Aronia melanocarpa L. berries (AME), characterized by their strong antioxidative potential
Agnieszka Białek et al.
Antioxidants (Basel, Switzerland), 9(3) (2020-03-21)
The aim of this study was to present overall lipid profile of organisms with ongoing neoplastic process and applied diet supplementation with pomegranate seed oil (PSO) and bitter melon extract (BME). The following were quantified in serum and cancerous tissues
Graziana Difonzo et al.
Foods (Basel, Switzerland), 8(4) (2019-04-26)
The shelf-life extension implicates the reduction of food waste. Plant polyphenols can have a crucial role in the shelf-life extension of foods. Olive leaf extract (OLE) is rich in phenolic compounds such as oleuropein, which is well-known for its antioxidant

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service