Skip to Content
Merck
All Photos(1)

Key Documents

450960

Sigma-Aldrich

Lutetium(III) chloride

anhydrous, powder, 99.99% trace metals basis

Synonym(s):

Lutetium chloride, Lutetium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LuCl3
CAS Number:
Molecular Weight:
281.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

99.99% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: lutetium

impurities

≤150.0 ppm Trace Rare Earth Analysis

mp

905 °C (lit.)

density

3.98 g/mL at 25 °C (lit.)

SMILES string

Cl[Lu](Cl)Cl

InChI

1S/3ClH.Lu/h3*1H;/q;;;+3/p-3

InChI key

AEDROEGYZIARPU-UHFFFAOYSA-K

Application

Lutetium(III) chloride can be used:
  • As a precursor to synthesize chiral bridged lutetium catalyst for asymmetric hydroamination.
  • As a starting material to prepare doped lutetium oxide nanophosphors.
  • To fabricate scintillation detectors such as lutetium aluminum garnet (LuAG).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjay Kumar Saxena et al.
Cancer biotherapy & radiopharmaceuticals, 32(10), 364-370 (2017-12-22)
The primary objective of this investigation is the development of a strategy for the synthesis of polyhydroxamic acid (PHA)-grafted cellulose film, its characterization, and evaluation of its usefulness for the preparation of
Development of new technologies for the manufacturing of nanocrystalline scintillation materials
Nikoloz V Jalabadze, et al.
IEEE transactions on nuclear science, 55, 1514-1522 (2008)
A Chiral-Bridged Aminotroponiminate Complex of Lutetium as Catalyst for the Asymmetric Hydroamination
Nils Meyer, et al.
Organometallics, 25, 4179-4182 (2006)
Usha Pandey et al.
Cancer biotherapy & radiopharmaceuticals, 34(5), 306-315 (2019-06-13)
Background: Transarterial radioembolization (TARE) represents an effective targeted therapeutic option for hepatocellular carcinoma (HCC), a cancer with high mortality
Preparation of nanocrystalline Lu2O3:Eu phosphor via a molten salts route
J. Trojan-Piegza and E. Zych
Journal of alloys and compounds, 380, 118-122 (2004)

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service