Skip to Content
Merck
All Photos(2)

Key Documents

450235

Sigma-Aldrich

Potassium gold(III) chloride

99.995% trace metals basis

Synonym(s):

Potassium tetrachloroaurate(III), Gold potassium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KAuCl4
CAS Number:
Molecular Weight:
377.88
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder
solubility:
H2O: soluble
alcohol: soluble
ethanol: soluble

grade

for analytical purposes

Assay

99.995% trace metals basis

form

powder

impurities

≤55.0 ppm Trace Metal Analysis

solubility

H2O: soluble
alcohol: soluble
ethanol: soluble

SMILES string

[K+].Cl[Au-](Cl)(Cl)Cl

InChI

1S/Au.4ClH.K/h;4*1H;/q+3;;;;;+1/p-4

InChI key

JMONCQYBAMKVQQ-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

General description

The yellow crystals of potassium gold chloride are soluble in water. 1,2

Application

Key raw material for synthesis of new gold(III) dithiolate complexes for luminescence studies or possible uses as photosensitizers or photocatalysts.1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M. Adnan Mansour et al.
Inorganic chemistry, 37(18), 4625-4632 (2001-10-24)
Two Au(III) dithiolate complexes, [Au(dbbpy)(tdt)]PF(6) and Au(eta(2)-C,N-ppy)(tdt), (dbbpy = 4,4'-di-tert-butyl-2,2'-bipyridine; tdt = 3,4-toluenedithiolate; ppy = C-deprotonated 2-phenylpyridine), have been prepared and structurally characterized by X-ray crystallography. The complexes have low-energy absorption bands that exhibit mild solvatochromism (lambda(max) = 444 nm
Andrew R Garcia et al.
Colloids and surfaces. B, Biointerfaces, 105, 167-172 (2013-02-05)
Fluorescent insulin fibrils gold nanoclusters (Au NCs) have been synthesized through the reduction of gold by human insulin in fibrillated form. Likewise, nanocluster formation has been regulated by insulin, working as a protein-based template. Environment- and surface-controlled experiments have shown
S Veeraapandian et al.
Journal of biomedical nanotechnology, 8(1), 140-148 (2012-04-21)
Escherichia coli is used for eco-friendly extra cellular synthesis of metallic silver and gold nanoparticles. This is achieved by reducing AgNO3 and HAuCl4 solutions respectively under ambient conditions by using the extra cellular protein produced by the microorganism. The proteins
Xiaorong Zhang et al.
Nanotechnology, 22(35), 355603-355603 (2011-08-11)
In this paper, a pH-inductive protein-scaffold biosynthesis of shape-tunable crystalline gold nanoparticles at room temperature has been developed. By simple manipulation of the reaction solution's pH, anisotropic gold nanoparticles including spheres, triangles and cubes could be produced by incubating an
Sergey S Zalesskiy et al.
Journal of the American Chemical Society, 135(9), 3550-3559 (2013-01-02)
Soluble gold precatalysts, aimed for homogeneous catalysis, under certain conditions may form nanoparticles, which dramatically change the mechanism and initiate different chemistry. The present study addresses the question of designing gold catalysts, taking into account possible interconversions and contamination at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service