Skip to Content
Merck
All Photos(3)

Key Documents

334049

Sigma-Aldrich

Gold(III) chloride

99%

Synonym(s):

Trichlorogold

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AuCl3
CAS Number:
Molecular Weight:
303.33
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

Assay

99%

form

powder

reaction suitability

reagent type: catalyst
core: gold

SMILES string

Cl[Au](Cl)Cl

InChI

1S/Au.3ClH/h;3*1H/q+3;;;/p-3

InChI key

RJHLTVSLYWWTEF-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gold(III) chloride is a Lewis acid with monoclinic structure. It is a hygroscopic and light-sensitive solid. It is frequently utilized to dope organic conducting compounds, to boost their conductivity. It also serves as a catalyst in organic synthesis and a precursor to synthesize gold nanoparticles for various electrochemical applications.

Application

Gold(III) chloride can be used:
  • As a dopant to fabricate graphene protection layer between PEDOT:PSS and ITO for perovskite solar cells. It helps to enhance the photoconversion efficiency by protection ITO and collecting holes.
  • To fabricate flexible and transparent electrodes for OLEDs. It fills the defects in graphene and improves current efficiency without affecting the transmittance.
  • As a precursor for green synthesis of gold nanoparticles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ligand-induced gold nanocrystal superlattice formation in colloidal solution.
Lin XM, et al.
Chemistry of Materials, 11(2), 198-202 (1999)
A comparative study of galvanic replacement reactions involving Ag nanocubes and AuCl2? or AuCl4?
Au L, et al.
Advanced Materials, 20(13), 2517-2517 (2008)
Synthesis and optical properties of cubic gold nanoframes
Au L, et al.
Nano Research, 1(6), 441-449 (2008)
E Castro-Longoria et al.
Colloids and surfaces. B, Biointerfaces, 83(1), 42-48 (2010-11-23)
The development of production processes that can reduce the environmental impact, offer waste reduction and increase energy efficiency is an important step in the field of application of nanotechnology. In this work the filamentous fungus Neurospora crassa was screened and
Changerath Radhakumary et al.
The Analyst, 137(22), 5387-5391 (2012-10-06)
This article reports a simple dipping method for the detection of cyanide ions in water and biological samples, such as blood, using chitosan-gold nanoparticle (CH-Au NP) composite films prepared using CH-Au NP colloids. Here, gold nanoparticles were generated by heating

Articles

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

Noble-metal nanostructures find diverse applications from catalysis to biomedical research, leveraging surface properties in various fields.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service