Skip to Content
Merck
All Photos(2)

Key Documents

444464

Sigma-Aldrich

Polyanetholesulfonic acid sodium salt

Synonym(s):

Sodium polyanetholesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

mol wt

average Mv 9,000-11,000

SMILES string

C/C=C/C1=CC=C(C=C1)OC.[O-]S(=O)[O-].[Na+]

InChI

1S/C10H12O.Na.H2O3S/c1-3-4-9-5-7-10(11-2)8-6-9;;1-4(2)3/h3-8H,1-2H3;;(H2,1,2,3)/q;+1;/p-2/b4-3+;;

InChI key

JKJBFNAERWARKW-CZEFNJPISA-L

Looking for similar products? Visit Product Comparison Guide

General description

Canonical SMILES: CC=CC1=CC=C(C=C1)OC.[O-]S(=O)[O-].[Na+]

Application

Anticoagulant

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B L Wasilauskas et al.
Journal of clinical microbiology, 35(3), 588-590 (1997-03-01)
We previously reported an inhibitory effect on Mycobacterium avium-M. intracellulare (MAI) when blood collected and processed with the Isolator system was placed in BACTEC 12B bottles for radiometric monitoring. We sought to identify the specific component(s) of the Isolator lysis-anticoagulant
E R Schnur et al.
Journal of clinical microbiology, 27(4), 654-656 (1989-04-01)
During an 8-month period at Children's Hospital, Oakland, Calif., a 9% rate for positive blood culture for children with Neisseria meningitidis meningitis was identified. The blood culture system used in each case was the BACTEC NR 730. This rate seemed
Kerry Varettas et al.
Journal of clinical microbiology, 43(9), 4613-4615 (2005-09-08)
The tube coagulase test (TCT) performed directly from positive blood culture bottles has been used to reduce the turnaround time for identifying Staphylococcus aureus. Most reports have shown the test to be specific but often lacking sufficient sensitivity to be
A Stelzner et al.
Allergie und Immunologie, 32(2), 129-133 (1986-01-01)
The influence of Liquoid (0.00005, 0.00001%), Carrageenan (0.001% up to 0.000001%), Colchicine (0.01% up to 0.0025%), Indomethacin (1 X 10(-4) up to 2.5 X 10(-5) mol/1), Ethanol (10% up to 1.25%), Galactose (1 X 10(-1) up to 1 X 10(-2)
J Cerda et al.
British journal of experimental pathology, 67(5), 623-628 (1986-10-01)
Levels of plasma fibronectin (Fn) were 63% lower than normal 15 min after the intravenous injection of liquoid (P less than 0.01); 3 h later they were still low but rebounded to 35% above normal (P less than 0.01) by

Articles

Recently, layer-by-layer (LbL) assembly has emerged as a versatile, gentle and, simple method for immobilization of functional molecules in an easily controllable thin film morphology.1,2 In this short review, we introduce recent advances in functional systems fabricated by using the mild, yet adaptable LbL technique.

We present an article that discusses two applications in particular; first, using these layers as polyelectrolyte membranes to control permeability.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service