All Photos(2)
About This Item
Linear Formula:
ClSO2C6H4CO2H
CAS Number:
Molecular Weight:
220.63
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
95%
mp
128-130 °C (lit.)
functional group
carboxylic acid
SMILES string
OC(=O)c1cccc(c1)S(Cl)(=O)=O
InChI
1S/C7H5ClO4S/c8-13(11,12)6-3-1-2-5(4-6)7(9)10/h1-4H,(H,9,10)
InChI key
LMRKXSDOAFUINK-UHFFFAOYSA-N
Related Categories
General description
3-(Chlorosulfonyl)benzoic acid is a sulfonyl halide. It participates in the synthesis of novel anthranilic acid class of PPARδ (Peroxisome proliferator-activated receptor δ) partial agonists.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Barry G Shearer et al.
Bioorganic & medicinal chemistry letters, 18(18), 5018-5022 (2008-08-30)
Anthranilic acid GW9371 was identified as a novel class of PPARdelta partial agonist through high-throughput screening. The design and synthesis of SAR analogues is described. GSK1115 and GSK7227 show potent partial agonism of the PPARdelta target genes CPT1a and PDK4
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service