Skip to Content
Merck
All Photos(1)

Key Documents

440841

Sigma-Aldrich

(1R,2S)-(+)-cis-1-Amino-2-indanol

99%

Synonym(s):

(1R,2S)-(+)-cis-1-Amino-2-hydroxyindane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
Beilstein:
2803743
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

optical activity

[α]22/D +63°, c = 0.2 in chloroform

optical purity

ee: 99% (GLC)

mp

118-121 °C (lit.)

functional group

hydroxyl

SMILES string

N[C@H]1[C@@H](O)Cc2ccccc12

InChI

1S/C9H11NO/c10-9-7-4-2-1-3-6(7)5-8(9)11/h1-4,8-9,11H,5,10H2/t8-,9+/m0/s1

InChI key

LOPKSXMQWBYUOI-DTWKUNHWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

This cis-aminoindanol and its antipode have been used in the preparation of a series of potent HIV-1 protease inhibitory peptides. Also, they have served as chiral ligands in the catalytic asymmetric reduction of prochiral ketones with boranes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thompson, W.J. et al.
Tetrahedron Letters, 33, 2957-2957 (1992)
Dorsey, B.D. et al.
Tetrahedron Letters, 34, 1851-1851 (1993)
D'Aniello, F. et al.
The Journal of Organic Chemistry, 59, 3762-3762 (1994)
Askin, D. et al.
The Journal of Organic Chemistry, 57, 2771-2771 (1992)
W J Thompson et al.
Journal of medicinal chemistry, 35(10), 1685-1701 (1992-05-15)
By tethering of a polar hydrophilic group to the P1 or P1' substituent of a Phe-based hydroxyethylene isostere, the antiviral potency of a series of HIV protease inhibitors was improved. The optimum enhancement of anti-HIV activity was observed with the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service